Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033228.3 | NP_001028400.2 | 4 | 787 | ||
GenBank mRNA | AK029105.1 | - | 4 | 703 | ||
AK035870.1 | - | 4 | 788 | |||
AK043769.1 | - | 4 | 726 | |||
AK137169.1 | - | 4 | 553 | |||
AK141204.1 | - | 4 | 755 | |||
BC014765.1 | - | 4 | 651 |