Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139294.5 | NP_647455.3 | 9 | 1243 | ||
GenBank mRNA | AJ276307.1 | - | 7 | 927 | ||
AK140431.1 | - | 9 | 1244 | |||
S79859.1 | - | 1 | 16 |