Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053009.3 | NP_443735.2 | 3 | 1361 | ||
GenBank mRNA | AK040976.1 | - | 2 | 242 | ||
AK147502.1 | - | 3 | 676 | |||
AK147560.1 | - | 3 | 1365 | |||
AK169477.1 | - | 3 | 262 | |||
U05342.1 | - | 1 | 46 |