Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148934.2 | NP_683736.1 | 2 | 1026 | ||
GenBank mRNA | AK153467.1 | - | 2 | 953 |