Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145588.1 | NP_663563.1 | 7 | 1078 | ||
GenBank mRNA | AK044826.1 | - | 3 | 2953 | ||
AK075922.1 | - | 7 | 1069 | |||
AK084310.1 | - | 7 | 1066 | |||
AK088033.1 | - | 4 | 654 | |||
BC003427.1 | - | 7 | 1078 | 2615715 |