Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 9 | 1149 | ||
GenBank mRNA | AK083258.1 | - | 9 | 1142 | ||
AK087972.1 | - | 9 | 1107 | |||
AK220294.1 | - | 9 | 1091 | |||
BC015278.1 | - | 8 | 722 | 4221290 |