Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025337.3 | NP_079613.3 | 2, 3 | 520 | ||
GenBank mRNA | AF525358.2 | - | 2, 3 | 422 | ||
AJ271800.1 | - | 2, 3 | 437 | |||
AJ271801.1 | - | 2, 3 | 437 | |||
AK003915.1 | - | 2, 3 | 507 | |||
AK143203.1 | - | 2, 3 | 519 | |||
BC031857.1 | - | 2, 3 | 497 | 5124841 |