Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081412.2 | NP_001074881.1 | 19 | 3323 | ||
GenBank mRNA | AK017213.1 | - | 4 | 382 | ||
AK129482.1 | - | 16 | 1513 | |||
BC002193.1 | - | 2 | 118 | |||
BC048842.1 | - | 7 | 567 | |||
BC060270.1 | - | 19 | 2561 | |||
BC131682.1 | - | 20 | 2728 | 40111128 |