Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031867.2 | NP_114073.1 | 6 | 2371 | ||
GenBank mRNA | AF301161.1 | - | 6 | 2267 | ||
AF301162.1 | - | 6 | 2267 | |||
AF337040.1 | - | 6 | 2449 | |||
AK153579.1 | - | 6 | 2321 | |||
AY032622.1 | - | 6 | 2267 | |||
BC070448.1 | - | 5 | 4284 |
Set Membership: |
G-protein coupled receptor |