Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133882.2 | NP_598643.1 | 6 | 850 | ||
GenBank mRNA | AB077304.1 | - | 6 | 772 | ||
AB077305.1 | - | 6 | 772 | |||
AK050313.1 | - | 6 | 851 | |||
BC022129.1 | - | 4 | 481 | |||
BC096382.1 | - | 6 | 824 | 5102825 |