Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024236.2 | NP_077198.1 | 7 | 768 | ||
GenBank mRNA | AK011351.1 | - | 7 | 697 | ||
AK078123.1 | - | 7 | 721 | |||
AK159107.1 | - | 7 | 712 | |||
AK161660.1 | - | 7 | 661 | |||
AK166694.1 | - | 7 | 722 | |||
AK214053.1 | - | 5 | 378 | |||
BC002107.1 | - | 7 | 708 | 3487104 |