Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199011.1 | NP_950176.1 | 9 | 1064 | ||
GenBank mRNA | AK043933.1 | - | 914 | |||
AK048764.1 | - | 9 | 1106 | |||
AK132151.1 | - | 9 | 1061 | |||
AK170840.1 | - | 8 | 1160 | |||
BC062929.1 | - | 9 | 1064 | 6855058 | ||
BC066012.1 | - | 9 | 1041 |