Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080639.3 | NP_542370.3 | 2 | 294 | ||
GenBank mRNA | AF282730.1 | - | 2 | 225 | ||
AK038820.1 | - | 2 | 293 | |||
AK040814.1 | - | 2 | 293 | |||
AK079347.1 | - | 2 | 295 | |||
AK140367.1 | - | 2 | 294 | |||
BC064046.1 | - | 2 | 274 | 875422 |