Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026695.3 | NP_080971.2 | 5 | 561 | ||
GenBank mRNA | AA207939.1 | - | 5 | 444 | ||
AK002407.1 | - | 5 | 503 | |||
AK013284.1 | - | 5 | 525 | |||
AK150293.1 | - | 5 | 504 | |||
AK192361.1 | - | 1 | 52 | |||
BC049237.1 | - | 5 | 490 | 4241552 | ||
BC069877.1 | - | 5 | 520 | 5370580 |