Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053208.4 | NP_444438.2 | 3 | 1336 | ||
GenBank mRNA | AF340231.1 | - | 2 | 1078 | ||
AF453879.1 | - | 3 | 1258 | |||
AJ310547.1 | - | 3 | 1307 | |||
AK075582.1 | - | 3 | 1288 | |||
AK152056.1 | - | 3 | 1297 | |||
AK152223.1 | - | 3 | 1294 | |||
AK159206.1 | - | 3 | 1289 | |||
AK166661.1 | - | 4 | 1202 | |||
AK167477.1 | - | 3 | 1289 | |||
BC023299.1 | - | 3 | 1270 | 5135579 | ||
BC051436.1 | - | 2 | 1054 | 6312638 | ||
BC086764.1 | - | 3 | 1275 | 5685311 |