Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000430.3 | NP_000421.1 | 8 | 1358 | ||
XM_011523901.1 | XP_011522203.1 | 9 | 1407 | |||
XM_011523902.2 | XP_011522204.1 | 9 | 1239 | |||
XM_011523903.2 | XP_011522205.1 | 9 | 1157 | |||
XM_017024699.1 | XP_016880188.1 | 8 | 1108 | |||
XM_017024700.1 | XP_016880189.1 | 8 | 1185 | |||
XM_017024701.1 | XP_016880190.1 | 8 | 1101 | |||
XM_017024702.1 | XP_016880191.1 | 7 | 946 | |||
GenBank mRNA | AF208835.1 | - | 7 | 791 | ||
AF208836.1 | - | 7 | 791 | |||
AF208837.1 | - | 7 | 790 | |||
AF208838.1 | - | 7 | 790 | |||
AF400434.1 | - | 7 | 983 | |||
AK293918.1 | - | 3 | 286 | |||
AK303015.1 | - | 4 | 598 | |||
AK313078.1 | - | 8 | 1012 | |||
BC064638.1 | - | 8 | 1000 | 5786560 | ||
L13385.1 | - | 8 | 1007 | |||
L13386.1 | - | 8 | 1243 | |||
L13387.1 | - | 1226 | ||||
L25107.1 | - | 438 | ||||
S81396.1 | - | 4 | 436 |