Precision Water Bath PDF
Precision redefined
Thermo Scientific Precision water baths are setting a new standard with enhanced features that simplify workflows and maximize productivity. Their rugged construction, compact footprint, advanced temperature controls, ease of use, and safety features clearly make our water baths a smart choice for your smart lab.
Browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Thermo Scientific Precision water baths are:
  • Compact—Save valuable bench space with the small footprint of our general purpose baths.
  • Effortless—Automatically preheat and turn off your bath for efficient work scheduling with new auto-on and auto-off timers.
  • Intuitive—Simplify parameter setting and monitoring with new icon-based controller interface to help ensure safe operation.
  • Safe—Protect your work with features such as audible alarm,  low-level detection and high-temperature cut-offs.
  • Everywhere—Access worldwide service and support when you need it for added peace of mind.
Download brochure
Featured precision water baths
Applications
Thermo Scientific Precision Water Baths
 General PurposeCirculatingColiformShakingDubnoff
Bacteriological  Examinations•••••
Coagulation Tests•• • 
Coliform  Determinations ••• 
Copper Strip Corrosion Tests••  •
Crude Oil Studies••   
Cytochemistry••  •
Dialysis    •
Demulsibility Studies••   
Enzyme Studies •  •
Electrophoresis Gel Destaining   • 
Environmental Studies•••••
Food Processing QC•••• 
Genetic Studies•• ••
Hormone Studies•• ••
Immunological Research•• ••
Incubation for Microbiological Assays•••• 
Incubation for Microcentrifugation Tubes•   •
Melting Agar•    
Metallurgical Analysis•• ••
Molecular Biology•• ••
Protein Analysis••   
Radioactive Isotope Uptake Studies•• ••
Radiochemistry•• • 
Serological Research•• ••
Thawing•• ••
Thawing Cryopreservation Vials•    
Tissue Culture Research•• ••
Virology Research•• ••
Warming Reagents••   
Water Quality Research•••••