SwiftArrayStudio Microarray Analyzer Arrays
Seamless assay compatibility—powered by SwiftArrayStudio
Human, agrigenomics and custom-built assays are designed to integrate seamlessly with SwiftArrayStudio automated array processing, including hybridization, washing, staining, and scanning—helping reduce manual handling after target preparation and supporting reproducible, high-throughput research workflows.
Human genotyping arrays
PangenomePro genotyping array
Population genomics, biobanking, and polygenic risk score research
PangenomePro arrays are designed to support large-scale human genomics research across diverse populations, with broad genome-wide coverage and content optimized for population-level analyses.
Key content highlights
  • ~2 million markers per array (approximate), providing dense genome-wide coverage
  • Common and rare variant content informed by global population datasets
  • Ancestry-aware design, supporting studies across diverse and admixed populations
  • Optimized for polygenic risk score (PRS) research, enabling the calculation and evaluation of genetic risk models in research settings
  • Content suitable for:
    • Genome-wide association studies (GWAS)
    • Population genomics and biobanking studies
    • Discovery-driven trait and risk research
Research applications supported
  • Population-scale variant discovery
  • Genetic architecture of complex traits
  • Polygenic risk score development and evaluation
  • Cross-population comparative genomics research
PharmacoPro genotyping array
Pharmacogenomics research and drug response studies
PharmacoPro arrays are designed for pharmacogenomics research, with curated content focused on genetic variation relevant to drug metabolism, transport, and response—strictly for research use.
Key content highlights
  • Targeted coverage of pharmacogenomics-relevant genes and pathways, including:
    • Drug-metabolizing enzymes
    • Drug transporters
    • Genes involved in pharmacokinetics and pharmacodynamics
  • Content informed by known PGx research markers across multiple therapeutic research areas
  • Designed to support:
    • Drug response research
    • Genetic variability studies in pharmacology
    • Research into genotype-drug associations
Research applications supported
  • Pharmacogenomics research studies
  • Population-level drug response analysis
  • Research into variability in drug metabolism and transport
Agrigenomics genotyping arrays
Veterinary and agricultural genomics research
SwiftArrayStudio supports agrigenomic research using Applied Biosystems single- and multi-species genotyping arrays for diploid species. Researchers, breeders, service providers and large enterprises can investigate genetic variation, genomic selection, parentage, breeding strategies, and economically important traits using the rapid SwiftArray workflow and automated array processing.
Key applications
  • Genome-wide SNP analysis
  • Genomic selection and breeding research
  • Parentage verification and traceability
  • Trait discovery and marker-assisted selection
  • Population genetics and diversity studies
  • Copy number variation (CNV) analysis on supported arrays
  • Agriculture genomics
Research applications include:
  • Livestock genomics
  • Aquaculture genomics
  • Companion animal genomics
  • Applied Biosystems Axiom expert diploid arrays (384F)
  • Custom single- and multi-species diploid arrays through the Axiom myDesign program in 96 or 384 format
Custom genotyping arrays
Tailored content for specialized research studies
For research groups with specific study requirements, custom Axiom arrays enable flexible assay design using validated Axiom probe chemistry.
Key capabilities
  • Available in 24-, 96-, mini-96, and 384-array formats
  • Custom content design supported through our Microarray Research Services Lab (MRSL) myDesign arrays
  • Suitable for:
    • Targeted variant panels
    • Follow-up studies and replication cohorts
    • Hypothesis-driven research designs
    • Multi-species designs
Custom arrays allow researchers to focus on variants, regions, or pathways most relevant to their scientific questions—while leveraging the trusted Axiom array workflow on the SwiftArrayStudio Analyzer.
For Research Use Only. Not for use in diagnostic procedures.