Original Slide-A-Lyzer Dialysis Cassettes Discontinuation
As part of our commitment to innovation and excellence, we are discontinuing a part of our current Slide-A-Lyzer product line to make way for the advanced G3 Slide-A-Lyzer cassettes that offer remarkable benefits for your laboratory needs.
  • Self-flotation—no external flotation devices or clips needed
  • Redesigned cap—secure locking cap with a wide opening for easy sample access
  • Green certified—uses 23–68% less plastic, helping support sustainability
  • Multiple sizes and MWCOs—available in 2K, 3.5K, 10K, and 20K MWCOs for 1–25 mL samples
  • Sterile option—E-beam-irradiated 10K MWCO cassettes available
  • Compact design—saves valuable lab space
Tentative effective date: March 31st, 2026
Upgrade to Slide-A-Lyzer G3 Dialysis Cassettes and elevate your research. If you're interested in a sample of the new Slide-A-Lyzer G3 dialysis cassettes, please contact your local sales representative.
Please see the table below for a list of affected products and their substitutes.
Affected products and their substitutes
Slide-A-Lyzer Dialysis Cassettes MWCO 2K
Discontinued product
Volume
Replacement product
Volume
Quantity
Catalog #
66205​
0.5 mL
Slide-A-Lyzer MINI Dialysis Devices
0.1 mL
50 devices*
69580
66203​
3 mL
Slide-A-Lyzer G3 Dialysis Cassettes
3 mL
10 cassettes
A52961
66212​
12 mL
Slide-A-Lyzer G3 Dialysis Cassettes
15 mL
8 cassettes
A52962
66230​
30 mL
Slide-A-Lyzer G3 Dialysis Cassettes
30 mL
6 cassettes
A52963​ 
New size
Slide-A-Lyzer G3 Dialysis Cassettes
125 mL
6 cassettes
A52965
*Not an equivalent alternative. The Slide-A-Lyzer MINI Dialysis Device 10K MWCO, 0.1 mL (Cat # 69580) are suitable for 10–100 µL samples, while the Slide-A-Lyzer G1 Dialysis Cassettes 2K MWCO, 0.5 mL (Cat # 66205) accommodates 100–500 µL sample volumes.
Slide-A-Lyzer Dialysis Cassettes MWCO 3.5K
Discontinued product
Volume
Replacement product
Volume
Quantity
Catalog #
66333​
0.5 mL
Slide-A-Lyzer MINI Dialysis Devices
0.5 mL
25 devices
88400
66330​
3 mL
Slide-A-Lyzer G3 Dialysis Cassettes
3 mL
10 cassettes
A52966
66110​
12 mL
Slide-A-Lyzer G3 Dialysis Cassettes
15 mL
8 cassettes
A52967
66130​
30 mL
Slide-A-Lyzer G3 Dialysis Cassettes
30 mL
6 cassettes
A52968 
New size
Slide-A-Lyzer G3 Dialysis Cassettes
125 mL
6 cassettes
A52970
Slide-A-Lyzer Dialysis Cassettes MWCO 7K
Discontinued product
Volume
No equivalent alternative available.
Use the 3.5K MWCO G3 Slide-A-Lyzer and/or MINI Dialysis Devices replacement products listed above.
66373​
0.5 mL
66370​
3 mL
66710​
12 mL
Slide-A-Lyzer Dialysis Cassettes MWCO 10K
Discontinued product
Volume
Replacement product
Volume
Quantity
Catalog #
66383​
0.5 mL
Slide-A-Lyzer MINI Dialysis Devices
0.5 mL
25 devices
88401
66380​
3 mL
Slide-A-Lyzer G3 Dialysis Cassettes
3 mL
10 cassettes
A52971
66810​
12 mL
Slide-A-Lyzer G3 Dialysis Cassettes
15 mL
8 cassettes
A52972
66830​
30 mL
Slide-A-Lyzer G3 Dialysis Cassettes
30 mL
6 cassettes
A52973​ 
New size
Slide-A-Lyzer G3 Dialysis Cassettes
125 mL
6 cassettes
A52975
66454​
0.5 mL
No equivalent alternative available.​
66455​
3 mL
Slide-A-Lyzer G3 Dialysis Cassettes
E-Beam Irradiated
3 mL
10 cassettes
A52981
66453​
12 mL
Slide-A-Lyzer G3 Dialysis Cassettes
E-Beam Irradiated
15 mL
8 cassettes
A52982
66456​
30 mL
Slide-A-Lyzer G3 Dialysis Cassettes
E-Beam Irradiated
30 mL
6 cassettes
A52983​ 
New size
Slide-A-Lyzer G3 Dialysis Cassettes
E-Beam Irradiated
125 mL
6 cassettes
A52985
Slide-A-Lyzer Dialysis Cassettes MWCO 20K
Discontinued product
Volume
Replacement product
Volume
Quantity
Catalog #
66005​
0.5 mL
Slide-A-Lyzer MINI Dialysis Devices
0.5 mL
25 devices
88402
66003​
3 mL
Slide-A-Lyzer G3 Dialysis Cassettes
3 mL
10 cassettes
A52976
66012​
12 mL
Slide-A-Lyzer G3 Dialysis Cassettes
15 mL
8 cassettes
A52977 
66030​
30 mL
Slide-A-Lyzer G3 Dialysis Cassettes
30 mL
6 cassettes
A52978
New size
Slide-A-Lyzer G3 Dialysis Cassettes
125 mL
6 cassettes
A52980
Contact us
If you have any questions regarding the transition or if we can be of assistance in any way, please feel free to contact our customer service.
For Research Use Only. Not for use in diagnostic procedures.