Search
After thawing from liquid nitrogen, SW480 cells were maintained in Nunclon Delta T25 cell culture flasks in Gibco DMEM medium supplemented with 10% Gibco FBS and 1% Pen-Strep for 2 passages before seeding for spheroid generation. ATCC protocol was followed for subculturing.
| Cell number to seed | Amount of Collagen I per ml of media |
|---|---|
| 10,000 cells | 30 µg |
| 5,000 cells | 15 µg |
| 2,500 cells | 8 µg |
| 1,250 cells | 4 µg |
| 625 cells | 2 µg |
| 312 cells | 1 µg |
Legend: SW480 spheroids generated in Nunclon Sphera Plate in various densities from 625 to 10,000 cells. Images were captured at Day 2 and Day 7 using EVOS 7000 microscope. No ECM bar represents merely cell aggregate and not a spheroid, whereas with Collagen I spheroids were uniform and intact. Scale bar equals to 650 µm.
Each cell density tends to generate different spheroid sizes; hence it is important to assess the density of your spheroid before proceeding to high throughput experiments. The graph below projects the different spheroid sizes we obtained with respect to their cell density in SW480 cell line. A good spheroid size for HT assays would be around 300-500 µm of diameter.
Sphericity is another phenomenon that defines the roundness or circular nature of a spheroid. Greater sphericity ensures the spheroid is intact, circular, and hard to disintegrate.
For Research Use Only. Not for use in diagnostic procedures.