1-Step™ NBT/BCIP Substrate Solution - FAQs
View additional product information for 1-Step™ NBT/BCIP Substrate Solution - FAQs (34042)
4 product FAQs found
Will the 1-Step NBT/BCIP Substrate Solution work with PVDF membranes?
Yes, it will work with both PVDF as well as nitrocellulose membranes
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
With the 1-Step NBT/BCIP Substrate Solution, how can I eliminate endogenous phosphatase activity?
To eliminate endogenous phosphatase activity, add a final concentration of 1 mM levamisole to the 1-Step NBT/BCIP Substrate Solution.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use the 1-Step NBT/BCIP Substrate Solution for ELISA?
No, the 1-Step NBT/BCIP Substrate Solution cannot be used for ELISA. It is a precipitating alkaline phoshpatase substrate developed for immunoblotting, immunohistochemistry and in situ hybridzation. For ELISA, use our 1-Step PNPP Substrate Solution (Cat. No. 37621).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.