Reliable water jacket performance
Thermo Scientific Forma Water Jacketed CO2 Incubators combine precise CO2 control and offer a choice of advanced features combinations to provide outstanding temperature stability, and excellent parameter recovery characteristics. Water jacked CO2 incubators are a top choice for researchers in academic, clinical, biotech and R&D labs around the world.
Trustworthy water jacket technology
Water jacket technology triple-wall construction provides outstanding temperature stability supplied by dual layers of water and high-quality insulation. This supports excellent thermal stability, which protects cell cultures against ambient temperature swings and unexpected power outages that put cultures at risk. 
Temperature loss from a power outage or extreme ambient swings can harm cultures. Product testing during a power failure in an 18º C (64.4º F) ambient environment showed the water jacketed incubator’s temperature gradually dropping just 1º C, from 37º C to 36º C (98.6º F to 96.8º F), in 1 hour and 7.6º C in 10 hours.
Features that meet the demands of your workflow
A number of features and options are available depending on the model of Forma Water Jacked CO2 Incubator you choose – these include:
HEPA filter airflow system – HEPA continuously filters the entir chamber volume every minute to provide an aseptic culturing atmosphere. HEPA-VOC filters can be chosen for removal of volatile organic compounds found in lab solvents and cleaning agents
Relative Humidity sensors – Optional RH sensors will monitor current conditions, display them on the display, and alert if the water runs low. They will also assist in automatically compensating for the effect of RH on CO₂.
Removable water pan – The Water Jacketed 3 models offer exceptional relative humidity control with a removable water pan
Stackable design – Forma Water Jacketed CO2 Incubators can be stacked to increase culture volume within the same lab footprint
Remote monitoring - 4-20mA signal output is included for interfacing with external data collection systems such as the Thermo Scientific Smart-Vue Pro Remote Monitoring System which provides external sensors and CFR-21 compliant software packages suitable for GMP environments.
Compare features of the Forma Water Jacketed CO2 Incubators
Model name
Forma Series II Water-Jacketed CO2 Incubator Forma Series 3 Water Jacketed CO2 Incubator
Control system
Touch button
i-CAN touchscreen display with data logging
CO2 control
0-20 % at ±0.1%
0-20 % at ±0.1%
CO2 sensor technology
TC or IR Sensor options
TC or IR Sensor options
Temperature range
Ambient +5° to 50°C
Ambient +5° to 50°C
Relative humidity
<90% @ 37 °C
<90% @ 37 °C
HEPA filtration
Included
Included
Chamber dimensions W x H x D
66.0 cm x 100.3 cm x 63.5 cm,
26.0 in x 39.5 in x 25.0 in
66.0 cm x 100.3 cm x 63.5 cm,
26.0 in x 39.5 in x 25.0 in
Chamber volume
184 L
184 L
Stackable
Optional
Optional
Chamber material
Stainless steel. Optional 100% copper interior component kit
Stainless steel. Optional 100% copper interior component kit
Data monitoring and collection
4-20mA signal output
4-20mA signal output
USB
HEPA Filters and Accessories
Shop for replacement consumables for your Forma CO2 Incubators. For all other accessory items, speak with a specialist.
Catalog # Name Size Price Qty
760175 Each
98.90
Online Exclusive
115.00 Save 16.10 (14%)
1900067 Each
177.00
(Contact Us ›)
-
760199 Each -
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.