Search
Our kits and plates for use with in vitro diagnostic (IVD) real-time PCR systems are designed to help you calibrate and verify the performance of your system quickly and easily. We recommend that you calibrate and verify according to your instrument’s user manual.
Applied Biosystems Spectral Calibration Kits contain pure dye standards; the real-time PCR system collects the pure spectra information from the standards. The system's application algorithm then uses this calibration information to help ensure accurate data analysis.
The TaqMan RNase P Instrument Verification Plates are pre-loaded with real-time PCR reagents and the TaqMan Assay for the single-copy RNase P gene. Use the plates to verify the performance of your Applied Biosystems real-time PCR system used for IVD-platform.
Quick links: Applied Biosystems QuantStudio 5 Dx System | Applied Biosystems QuantStudio Dx System |
7500 Dx System
See user's manual or package insert for limited label license, and trademark information.
For In Vitro Diagnostic Use.