Get the most from your requirements for routine and high-throughput UV-Vis/Vis measurements. Built for demanding real-world applications, rugged and compact Thermo Scientific GENESYS spectrophotometers are ideal for walk-up measurements or high-throughput QC/QA testing, research labs, or the classroom.
Choose from UV-Vis or Vis configurations for single cuvette/test tube applications, quantitative analysis, multiple-cell changers, temperature control, remote sampling, double beam reference cell design, or pre-programmed life science methods. All instruments feature superior performance, sampling convenience, user-friendly operation and are built to withstand heavy use.
Need help choosing? Check out the GENESYS Selection Guide.
On a budget? See limited-time promotional offers.
Ready to try it out? Talk to an expert about a trial.
GENESYS 30 Vis Spectrophotometer
Outstanding durability for routine QA/QC labs with walk-up simplicity and a removable, washable sample compartment liner for easy clean up.
GENESYS 40 Vis/50 UV-Vis Spectrophotometer
Setting a new standard is single cell QA/QC analysis and support for a variety of cuvettes and test tubes, plus a sipper accessory and thermostatted cell holder.
GENESYS 140 Vis/150 UV-Vis Spectrophotometer
Research quality measurements with routine simplicity. Dual beam optics ensure accuracy across a range of options for sampling, data export and remote analysis.
GENESYS 180 UV-Vis Spectrophotometer
Double beam optical design to measure reference and sample simultaneously, plus all of the features of the GENESYS 150 spectrophotometer.
Quick links
Legendary reliability. Unrivaled usability.
Built on the foundation of 60 years of spectroscopy experience, the next generation Thermo Scientific GENESYS family of Vis / UV-Vis spectrophotometers provides exceptional performance with a contemporary design. There is a GENESYS spectrophotometer you can count on to provide accurate, reproducible results for your application requirements.