Anza™ DNA End Repair Kit
Anza™ DNA End Repair Kit
Invitrogen™
Anza™ DNA End Repair Kit
The Invitrogen™ Anza™ DNA End Repair Kit is used to convert DNA with cohesive ends to blunt-ended 5’-phosphorylated DNA forRead more
Have Questions?
Catalog NumberQuantity
IVGN250420 reactions
Catalog number IVGN2504
Price (USD)
198.00
Each
Add to cart
Quantity:
20 reactions
Price (USD)
198.00
Each
Add to cart
The Invitrogen™ Anza™ DNA End Repair Kit is used to convert DNA with cohesive ends to blunt-ended 5’-phosphorylated DNA for use in blunt-end ligation reactions. The included pre-mixed Anza™ DNA End Repair Mix contains T4 DNA polymerase, Klenow Fragment, and T4 PNK (polynucleotide kinase) to minimize pipetting, while the Anza™ 10X End Repair Buffer contains ATP and dNTPs to facilitate the activity of the enzyme mix.
Benefits:
Convenient—pre-mixed enzyme mixture for superior convenience and reduction of pipetting
Efficient—simultaneous blunting and phosphorylation
Compatible—use directly in Anza restriction enzyme digestion reaction mixture following heat inactivation
Fast—end repair complete in 15 minutes
The Anza DNA End Repair Kit is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza T4 PNK Kit, and Anza DNA Blunt End Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferAnza™ 10X End Repair Buffer
Product TypeDNA End Repair Kit
Quantity20 reactions
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymeT4 Polynucleotide Kinase, T4 DNA polymerase, Klenow Fragment
Product LineAnza™
Unit SizeEach
Contents & Storage
100 μL Anza DNA End Repair Mix
200 μL Anza 10X End Repair Buffer
Store at -5 to -30°C.