The KingFisher Flex is the most versatile benchtop automated extraction instrument in the lab for 24 or 96 samples per run. Consistent high-throughput extraction and purification of DNA, RNA, Protein, and Cells.
KingFisher Instruments for SARS-CoV-2 testing
KingFisher Flex features
- High-throughput purification of 24-96 samples per run
- Easy to install and run (ready to run in 10 minutes or less)
- Choose from two plate formats for wide volume range (20–5,000 µL)
- Download existing protocols from library, or easily customize protocols using Thermo Scientific BindIt Software
- Simple workflow
KingFisher Flex details
Instrument size | Benchtop |
Throughput level | Medium to high |
Processing volume range |
|
Samples per run | 24 or 96 |
Customizable protocols | Yes |
Heating/cooling | From 5°C above ambient temperature to 115°C |
Ultraviolet lamp | No |
* Or similar skirted PCR plate.
How KingFisher instruments work
KingFisher instruments automate extraction of DNA, RNA, protein, and cells by moving magnetic beads (not liquids), leading to clean extractions and enabling consistent results. Using a simple process (bind, wash, elute), KingFisher instruments can automate the extraction of any analyte of interest with a bead on it.
Kits and reagents
Choose from a wide range of magnetic bead-based kits and reagents from trusted brands, such as Dynabeads and MagMAX.
Plates and plastics
- Made of polypropylene
- Low binding affinity for biomolecules, ideal for magnetic particle processing
- Sterile plastics come with a purity certificate and are guaranteed to be free of RNase, DNase, DNA and endotoxin contamination, shelf life is 5 years
Select three different plate formats for a range of input volumes.
Application note: MagMAX Viral/Pathogen kit protocol changes to enable increased SARS-CoV-2 testing throughput
KingFisher plastics for SARS-CoV-2 testing
KingFisher Flex software and protocols
Nucleic Acid Protocols
- Protocols designed for nucleic acid purification procedures and reagent kits
IP/Protein Protocols
- Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
BindIt Software
- Create and store protocols in a PC database
- Compatible with all KingFisher instruments
- Easy to use
- Generate reports
Service and support
Instrument and application specialists
Whether servicing your KingFisher purification system or addressing your workflow application questions, our more than 1,000 trained professionals make up the industry’s largest network, ready to assist you when you need it.
Quick-start training
The KingFisher system includes SmartStart orientation to get you up and running quickly in your lab. The orientation includes basic familiarization and setup with online instrument management and includes on-site training with standard kits.
Warranty and service plans
Every KingFisher system comes with a one-year warranty. Extended-coverage service plans are also available at the time of instrument purchase. Whether your laboratory requires the highest service levels and adherence to stringent regulatory guidelines, or you need to maximize performance with a limited budget—or anything in between—we’ll work with you to develop a solution that best suits your needs. KingFisher instrument services and support
Find out more at thermofisher.com/instrumentservices or contact us at thermofisher.com/contactus.
KingFisher Flex how-to videos
How to unbox the KingFisher Flex
Guiding you through the process to safely unbox your KingFisher Flex Instrument.
Remove transportation locks
After receiving your KingFisher Flex, this video will walk through the steps to remove the transportation locks.
Install tip shield
Walking through the process to install the tip shield on the KingFisher Flex.
Power on
What to know in order to power on the KingFisher Flex Instrument.
Change heat block
Helping identify the process to change the heat block on the KingFisher Flex instrument.
Install magnet head and check tip comb
Helping with the steps to install the magnet head of the KingFisher Flex and also how to check the tip comb before loading instrument.
Connect computer and use BindIt software
Walking through the process to connect the KingFisher Flex top laptop and installing the BindIt software.
Transfer BindIt software protocols
Update your instrument with your protocol, simple steps to transfer your BindIt protocol to the KingFisher Flex instrument.
Remove magnet head
Simple steps to help remove the magnet head safely from the KingFisher Flex instrument.
Related products for plate setup
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
E1-ClipTip Equalizer Pipettes
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Multidrop Combi Reagent Dispenser
Automated reagent dispenser. Learn more
KingFisher Flex features
- High-throughput purification of 24-96 samples per run
- Easy to install and run (ready to run in 10 minutes or less)
- Choose from two plate formats for wide volume range (20–5,000 µL)
- Download existing protocols from library, or easily customize protocols using Thermo Scientific BindIt Software
- Simple workflow
KingFisher Flex details
Instrument size | Benchtop |
Throughput level | Medium to high |
Processing volume range |
|
Samples per run | 24 or 96 |
Customizable protocols | Yes |
Heating/cooling | From 5°C above ambient temperature to 115°C |
Ultraviolet lamp | No |
* Or similar skirted PCR plate.
How KingFisher instruments work
KingFisher instruments automate extraction of DNA, RNA, protein, and cells by moving magnetic beads (not liquids), leading to clean extractions and enabling consistent results. Using a simple process (bind, wash, elute), KingFisher instruments can automate the extraction of any analyte of interest with a bead on it.
Kits and reagents
Choose from a wide range of magnetic bead-based kits and reagents from trusted brands, such as Dynabeads and MagMAX.
Plates and plastics
- Made of polypropylene
- Low binding affinity for biomolecules, ideal for magnetic particle processing
- Sterile plastics come with a purity certificate and are guaranteed to be free of RNase, DNase, DNA and endotoxin contamination, shelf life is 5 years
Select three different plate formats for a range of input volumes.
Application note: MagMAX Viral/Pathogen kit protocol changes to enable increased SARS-CoV-2 testing throughput
KingFisher plastics for SARS-CoV-2 testing
KingFisher Flex software and protocols
Nucleic Acid Protocols
- Protocols designed for nucleic acid purification procedures and reagent kits
IP/Protein Protocols
- Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
BindIt Software
- Create and store protocols in a PC database
- Compatible with all KingFisher instruments
- Easy to use
- Generate reports
Service and support
Instrument and application specialists
Whether servicing your KingFisher purification system or addressing your workflow application questions, our more than 1,000 trained professionals make up the industry’s largest network, ready to assist you when you need it.
Quick-start training
The KingFisher system includes SmartStart orientation to get you up and running quickly in your lab. The orientation includes basic familiarization and setup with online instrument management and includes on-site training with standard kits.
Warranty and service plans
Every KingFisher system comes with a one-year warranty. Extended-coverage service plans are also available at the time of instrument purchase. Whether your laboratory requires the highest service levels and adherence to stringent regulatory guidelines, or you need to maximize performance with a limited budget—or anything in between—we’ll work with you to develop a solution that best suits your needs. KingFisher instrument services and support
Find out more at thermofisher.com/instrumentservices or contact us at thermofisher.com/contactus.
KingFisher Flex how-to videos
How to unbox the KingFisher Flex
Guiding you through the process to safely unbox your KingFisher Flex Instrument.
Remove transportation locks
After receiving your KingFisher Flex, this video will walk through the steps to remove the transportation locks.
Install tip shield
Walking through the process to install the tip shield on the KingFisher Flex.
Power on
What to know in order to power on the KingFisher Flex Instrument.
Change heat block
Helping identify the process to change the heat block on the KingFisher Flex instrument.
Install magnet head and check tip comb
Helping with the steps to install the magnet head of the KingFisher Flex and also how to check the tip comb before loading instrument.
Connect computer and use BindIt software
Walking through the process to connect the KingFisher Flex top laptop and installing the BindIt software.
Transfer BindIt software protocols
Update your instrument with your protocol, simple steps to transfer your BindIt protocol to the KingFisher Flex instrument.
Remove magnet head
Simple steps to help remove the magnet head safely from the KingFisher Flex instrument.
Related products for plate setup
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
E1-ClipTip Equalizer Pipettes
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Multidrop Combi Reagent Dispenser
Automated reagent dispenser. Learn more
KingFisher Flex ordering information
Resources
Brochure
Application Notes
Technical Notes
Smart Note
For Laboratory Use.