Drying under vacuum allows liquids to evaporate at temperatures below the regular boiling point. Benefits include:
- Faster drying, since liquids evaporate at lower temperatures.
- Dry thermic instable materials very gently, at low temperatures (great for pharma/food drying applications).
- Reduce capillary forces and completely dry intermediate or finished products (great for electronics).
- Dry under inert gas atmosphere, e.g. to avoid oxidation processes.
Offered with various temperature ranges and sizes, Thermo Scientific vacuum ovens are designed with safety, reliability and efficiency in mind—and offer features that tailor to your specific applications. Choose from several different product lines - from basic to high end.
Browse our complete portfolio or let one of our experts help you make a selection.
Good – better – best approach, with feature and control options.
Microprocessor control with overtemperature protection for safety, this multipurpose line covers most heating and drying applications under vacuum which require good temperature performance.
Select the oven that's best for your application
![]() | ![]() | ![]() | ![]() | |
Temperature range | Ambient +10/+15° to 200°/300°/400°C | Ambient +6° to 260°C | Ambient +10°/50° to 280°C | Ambient +5° to 220°C |
Available sizes | 25/53/128L (0.88/1.87/4.52 cu. ft.) | 18.6/42.5/127.4L (0.65/1.5/4.5 cu. ft.) | 18.4/42.4L (0.65/1.5 cu. ft.) | 12.5/19.8/65L (0.44/0.7/2.3 cu. ft.) |
Features | PID control, digital display, analog control with dial thermometer, overtemperature cut-out, inert gas connection (on large models) | PID control, adjustable overtemperature protection, inert gas connection | Electronic control, fixed overtemperature protection, on/off timer (large models only), inert gas connection | Analog control with dial thermometer, overtemperature cut-out, inert gas connection (on large models) |
Sample applications |
|
|
|
|
Resources
Support
- Unity Lab Services
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.