Search
Thermo Fisher Scientific has partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument features high-end manufacturing and superior pipetting performance at an affordable price.
With the Axiom genotyping starter kit, this instrument automates the target preparation steps for the Axiom 2.0 assay, including DNA amplification, fragmentation, purification, resuspension, hybridization preparation, and stain preparation for GeneTitan MC Instrument. With the CytoScan starter kit, it automates the post-PCR target preparation steps for the CytoScan cytogenetic microarray assay.
Includes additional equipment and parts necessary to set up NIMBUS Target Preparation Instrument for Axiom Genotyping Solution.
Includes additional hardware required to set up NIMBUS Target Preparation Instrument for the CytoScan assay.
The Biomek FXP Target Prep Express is an Thermo Fisher-specific configuration of the successful Biomek FXP platform. It enables superior positional accuracy and robustness. The intuitive software interfaces enable you to start up quickly and easily switch between methods. Have confidence with fully validated Thermo Fisher target preparation methods, including Axiom Genotyping Solution target preparation, GeneChip HT WT Plus, GeneChip HT 3’ IVT Plus, and GeneChip HT Pico methods. Contact us for more information.
Target preparation on the Nimbus instrument helps minimize run-to-run variability and the labor burden associated with complex manual pipetting, helping to improve test reproducibility and laboratory efficiency.
For Research Use Only. Not for use in diagnostic procedures.