Transitioning human embryonic stem cells (hESCs) to a defined, feeder- and serum-free system can be challenging—but with the right approach, it’s achievable and reliable. The following guide offers tips for adapting hESCs from feeder-dependent or feeder-free media to StemPro hESC SFM (used with Geltrex matrix), covering recommended preparatory conditions, gradual versus direct adaptation strategies, and tips to maintain pluripotency, colony morphology, and culture stability.
Why adaptation matters
Here are some key reasons why adaptation matters when transitioning hESCs to StemPro hESC SFM:
- Helps ensure maintenance of undifferentiated morphology and pluripotency when moving from feeder-dependent or feeder-free systems.
- Helps avoid spontaneous differentiation due to culture stress or sub-optimal conditions during medium change.
- Enables reliable scaling and expansion of hESCs, supporting downstream applications such as gene engineering or differentiation.
- Helps minimize variability by moving to a defined, serum- and feeder-free system, which reduces batch-to-batch and feeder-dependent inconsistencies.
- Helps preserve normal karyotype and genetic stability during long-term culture in the new medium system.
Pre-adaptation checklist
Starter culture quality
- Ensure cells are 70–80% confluent, predominantly undifferentiated.
- Remove any differentiated colonies manually.
Substrate and coating requirements
- Use Geltrex hESC-qualified matrix or other matrix.
- Prepare coating plates ahead of time and follow recommended dilutions.
Media and feeding frequency
- Change medium daily during adaptation.
- Monitor colony edges and density; over- or under-seeding can lead to differentiation.
Tips adapting hESCs to StemPro hESC SFM from other feeder-free medium
When adapting cells to StemPro hESC SFM from feeder-free parent culture such as MEF conditioned medium or other feeder-free medium:
- Make a frozen stock of cells in control medium (MEF conditioned medium or other feeder-free medium) prior to adaptation.
- Maintain a stock culture in control medium throughout the hESC adaptation to StemPro hESC SFM as a backup.
Tips adapting hESCs to StemPro hESC SFM from feeder-dependent medium
When adapting cells to StemPro hESC SFM from a feeder-dependent parent culture:
- Better results are obtained when the parent hESC culture has been maintained on either murine embryonic fibroblast (MEF) or Human Fibroblast Feeder (HFF) cells.
- We recommend 100% MEF conditioned medium on Geltrex matrix as a control for the adaptation protocols described below.
- Some residual feeders may be observed in the cultures during the adaptation process but in subsequent passages the inactivated feeder cells will be passaged out.
Adaptation strategies
Sequential adaptation is gentle on cells. By gradually increasing exposure to the new media environment, it reduces shock and allows cells to acclimate, leading to higher survival and better retention of normal morphology and pluripotency markers. This is recommended for sensitive cell lines or those with a history of differentiation/instability.
Direct adaptation is faster and simpler. It helps minimize time and labor and gets cells into the new medium as soon as possible. However, it can be stressful for cells, especially sensitive or “finicky” lines—sudden changes in nutrients, growth factors, matrix, and environment may trigger differentiation, cell death, or loss of pluripotency.
Hybrid (direct + partial sequential) adaptation gives a balance between speed and caution. Part of the culture goes directly to new medium to accelerate adaptation, while backup/parallel cultures ensure that if direct transition fails, cells can be rescued without losing the line. It also provides experimental redundancy, which is useful when dealing with valuable or hard-to-derive PSC lines.
Sequential adaptation protocol
- Passage 1: 75% control medium + 25% StemPro hESC SFM Complete Medium + Geltrex matrix
- Passage 2: 50% control medium + 50% StemPro hESC SFM Complete Medium + Geltrex matrix
- Passage 3: 25% control medium + 75% StemPro hESC SFM Complete Medium + Geltrex matrix
- Passage 4 & thereafter: 100% StemPro hESC SFM Complete Medium + Geltrex matrix
Note: If hESC are extremely finicky, a further level of caution can be taken by maintaining a culture in each prior passage medium while starting the next level of adaptation. For example, when passaging the 25/75 control medium/StemPro hESC SFM culture (as described above), hESC can be passaged into both 100% StemPro hESC SFM and 25/75 medium. If the 100% culture does poorly, adaptation can be resumed using the backup 25/75 culture.
Direct adaptation protocol
- Passage hESCs directly into StemPro hESC SFM Complete Medium + Geltrex matrix.
Note: A 1:2 split ratio is suggested for the first 3 passages.
Direct and partial sequential protocol
- Split cells into three plates (Plate 1, 2, and 3).
- At the first passage, seed Plate 1 directly into StemPro hESC SFM Complete Medium + Geltrex matrix, and Plates 2 and 3 into the feeder-free control medium + Geltrex matrix.
- On the next day, fluid-change Plate 2 with StemPro hESC SFM Complete Medium + Geltrex matrix that day and every day thereafter.
- For Plate 3, at the second passage, try plating these cells directly into StemPro hESC SFM Complete Medium + Geltrex matrix at 1:2 split ratio (as above).
Using Geltrex matrix: Thin gel method (non-gelling)
- Thaw Geltrex matrix as described on the product insert.
- Mix Geltrex matrix by slowly pipetting solution up and down; be careful not to introduce air bubbles. Dilute 1 mL of Geltrex matrix into 29 mL of pre-chilled (2 to 8°C) DMEM/F12 medium.
Notes:
- Empirical determination of the optimal coating concentration for your application may be required. Volumes can be adjusted accordingly.
- Most customers have seen that a dilution higher than 1:30 may be appropriate for their hESC line. Try anywhere from 1:30 to 1:100.
- Add a sufficient amount of diluted Geltrex solution to cover the entire growth surface area (1.5 mL for 35 mm dish, 3 mL for 60 mm dish).
- Coat the dish and place at 37°C for a minimum of 60 minutes.
- The coated dish is stable for two weeks when stored at 2 to 8°C and sealed with Parafilm.
Notes:
- Do not allow coated surface to dry out. It is critical to maintain a storage temperature of 2 to 8°C to avoid premature gelling.
- At time of use, it is recommended to place plates at room temperature for an hour before aspirating. Aspirate Geltrex coating and immediately plate cells in preequilibrated cell culture medium.
Ordering information
A1000701, A1569601, 10565042
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments