Protocols
Introduction
This protocol was developed for the MultiPROBEŒ II HT Liquid Handling System with a Gripper Integration Platform. Time required: 1 hr 20 min.
For more information on the protocol, see the RNAqueous-96 Instruction Manual (pdf).
Ordering Information
Catalog # Name Size Price (USD) Qty
AM1920 RNAqueous™-96 Total RNA Isolation Kit Each
1,180.00
AM7020 RNAlater™ Stabilization Solution, 100 mL Each
206.65
Online Exclusive
215.00
Save 8.35 (4%)
AM7021 RNAlater™ Stabilization Solution, 500 mL Each
585.65
Online Exclusive
602.00
Save 16.35 (3%)
AM9780 RNaseZap™ RNase Decontamination Solution, 250 mL Each
99.65
Online Exclusive
105.00
Save 5.35 (5%)
AM9782 RNaseZap™ RNase Decontamination Solution, 6 x 250 mL Each
441.65
Online Exclusive
467.00
Save 25.35 (5%)
AM9784 RNaseZap™ RNase Decontamination Solution, 4 L Each
919.65
Online Exclusive
962.00
Save 42.35 (4%)
AM1940 NorthernMax™ Kit Each
788.65
Online Exclusive
812.00
Save 23.35 (3%)
AM1946 NorthernMax™-Gly Kit Each
908.00
Protocol
  1. Add 150 µl Lysis Solution to each well.
  2. Add 150 µl 64% ethanol to each well.
  3. Mix and transfer to the 96-well filter plate.
  4. Apply vacuum suction (20 mm Hg) for 3 min (RNA remains bound to the filter).
  5. Add 400 µl Wash Solution A to each well and apply vacuum suction (20 mm Hg) for 2 min.
  6. Add 40 µl (100 units) DNase I.
  7. Incubate 20 min at room temperature.
  8. Add 400 µl Wash Solution B to each well and apply vacuum suction (20 mm Hg) for 2 min.
  9. Add 400 µl Wash Solution C/D to each well and apply vacuum suction (20 mm Hg) for 2 min.
  10. Add 400 µl 100% ethanol to each well and apply vacuum suction (20 mm Hg) for 2 min.
  11. Move 96-well filter plate to dry heat block covered with fresh paper towels and heat at 42­C for 3 min (this removes any residual wash solution).
  12. Move Spacer and Collection plates to the vacuum manifold. Place 96-well filter plate on top.
  13. Add 80 µl Elution Buffer to each well.
  14. Apply vacuum suction at 10 mm Hg for 30 sec then at 20 mm Hg for 2 min (RNA is transferred to collection plate).
  15. (optional) Manually check for fluid remaining on filter plate. If fluid is present, centrifuge the filter plate/collection assembly to recover residual sample.
TOP
LT038