Search
Isolate or deplete CD45+ cells directly from whole blood, buffy coat or MNC suspensions with Dynabeads CD45. For rapid and consistent results in protein or gene expression analysis, lyse the CD45+ cells while they are still attached to the beads and directly process for further molecular analysis. Enrich circulating tumor cells (non-hematopoietic) from MNC suspensions with Dynabeads CD45.
Principle of Isolation
Dynabeads are mixed with the cell sample in a tube. The Dynabeads will bind to the target cells during a short incubation, and then the bead-bound cells are separated by a magnet.
Description of Materials
Dynabeads CD45 are uniform, superparamagnetic polystyrene beads (4.5 μm diameter) coated with a monoclonal mouse IgG2a antibody specific for an epitope common to all known CD45 isoforms.
Materials Supplied
Additional Materials Required
Magnet (Dynal MPC™)
Dynabeads Washing Procedure
Dynabeads should be washed before use.
1. Resuspend the Dynabeads in the vial.
2. Transfer the desired volume of Dynabeads to a tube.
3. Add the same volume of Buffer 1, or at least 1 ml, and mix.
4. Place the tube in a magnet for 1 min and discard the supernatant.
5. Remove the tube from the magnet and resuspend the washed Dynabeads in the same volume of Buffer 1 as the initial volume of Dynabeads.
Sample Preparation
MNC preparation from whole blood, buffy coat or bone marrow.
DNase treatment of BM-MNC to remove interfering DNA.
BM-MNC should be frozen in the presence of 120 IU DNase solution per 1 ml cell suspension.
Critical Steps for Cell Isolation
Table 1. Volume of Dynabeads added per ml of cell sample. The volumes can be scaled up as required.
| Positive isolation or depletion | Enrichment of circulating tumor cells from MNC | |
|---|---|---|
| Sample volume* | 1 ml (107 cells/ml) | 1 ml (2 x 107 /ml) |
| Volume of Dynabeads | 100 μl | 250 μl |
| Total no. of cells processed per product | 5 x 108 cells | 5 x 108 cells |
* If the concentration of cells is increased, the volume of Dynabeads must be increased accordingly 2 x 108 cells
Enrichment of Tumor Cells
Before cell isolation or transferring the enriched cells to a new tube, pre-coat the tubes for 5 min using buffer with FCS (e.g. Buffer 2).
Detection of Tumor Cells
Enriched tumor cells can be detected and analyzed by immunocytochemistry (ICC).
Other Cellular Applications
Enriched tumor cells isolated with Dynabeads CD45 are bead-free, viable and can be grown in culture.
Cell Isolation (general)
Add Dynabeads to the prepared sample according to table 1.
i) Add 1 ml Buffer 1 per 1 x 107 Dynabeads.
ii) Place the tube in the magnet for 1 min and discard the supernatant.
Invitrogen Dynal AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Storage/Stability
This product is stable until the expiry date stated on the label when stored unopened at 2-8°C. Store opened vials at 2-8°C and avoid bacterial contamination. Keep Dynabeads in liquid suspension during storage and all handling steps, as drying will result in reduced performance. Resuspend well before use.
Warnings And Limitations
This product is for research use only. Not intended for any animal or human therapeutic or diagnostic use unless otherwise stated. Follow appropriate laboratory guidelines. This product contains 0.02% sodium azide as a preservative, which is cytotoxic.
Avoid pipetting by mouth!
Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. When disposing through plumbing drains, flush with large volumes of water to prevent azide build up. Certificate of Analysis (CoA) is available upon request.
For Research Use Only. Not for use in diagnostic procedures.