Technical Resources - Media Formulations
14040 - DPBS, calcium, magnesium
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0100.00.9009009
Magnesium Chloride (MgCl2-6H2O)203.0100.00.49261084
Potassium Chloride (KCl)75.0200.02.6666667
Potassium Phosphate monobasic (KH2PO4)136.0200.01.4705882
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.02160.08.059702
Verweis
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
Related Products:
14080 - DPBS, calcium, magnesium
Cat No: 14080048 , 14080055 , 14080089
21300 - DPBS, powder, calcium, magnesium
Cat No: 21300025 , 21300058 , 21300058 , 21300074