Thermo Scientific Lab Oven Models
Take a closer look at the features and specifications of Thermo Scientific Heratherm Heating and Drying ovens and Thermo Scientific Vacuutherm and Lindberg Blue/M Vacuum Ovens. 
Thermo Scientific Heratherm Heating and Drying Oven models
Heratherm Advanced Protocol Security Ovens
Safety features offer protection of your most precious samples
Heratherm Advanced Protocol Ovens
Ultimate accuracy and increased heating
Heratherm General Protocol Ovens
For everyday heating and drying applications
Precision Compact Ovens
Compact ovens for drying and baking applications
Temperature range  
50 – 330°C 50 – 330°C 50 – 250°C +5 – 210°C
Capacity  
61 – 170 L models 61 – 731 L models 65 – 744 L models 48 L models
Heating options  
Mechanical and gravity convection
Programmability  
10 automatically repeatable programs, each with up to 10 discrete steps.
Access port for remote monitoring.
10 automatically repeatable programs, each with up to 10 discrete steps   None
Ordering information      
All regions: models Americas: 120V 60Hz models
Europe, Africa, Asia: 230V 60Hz models
Japan: 208-240V 60Hz, 208-240V 60Hz
Americas: 120V 60Hz 
Europe, Africa, Asia: 230V 50/60Hz
Americas only: 120V
Thermo Scientific Vacuum Oven models
Vacutherm Vacuum Ovens
Achieve precise temperatures for simple or complex drying processes including heat sensitive materials
Lindberg/Blue M Vaccum Ovens
Designed for life science, industrial, and general lab applications requiring higher performance standards
Thermo Scientific Vacuum Ovens
For everyday use including drying, curing, vacuum embedding, and plating applications
Temperature range
Up to 400 °C 6 – 260 °C Ambient +10—220 °C
Capacity
25 – 128 L 18.6 – 42.5 L 12.5 – 65 L options
Heating options
Jacket heating or shelf conduction Jacket heating Jacket heating
Safety features
Overtemperature sensor protects oven and sample
Double-pane safety glass doors
Access port in rear wall for remote monitoring
Independent overtemperature safety system factory set with reset pushbutton Polycarbonate safety shield protects door glass
Ordering information    
View models View models View models
Oven accessories
Heratherm Heating and Drying Oven
Accessories
Vacutherm Vacuum Oven
Accessories
Lindberg Blue/M Vacuum Oven
Accessories
Available accessories
  • Perforated shelving
  • Wire mesh shelving
  • Additional shelves
  • Aluminum trays
  • Daily Program Timers
  • Digicon S Temperature controllers
  • Eurotherm 2404/P4 temperature controllers
  • Precision control valves
  • RS 232 computer interfaces for Kelvitron™ controllers
  • Support frames
  • Stainless-steel vacuum connection kits for tubing
  • Stainless-steel trays
  • Thermicon P temperature program controllers
  • Vacuum chambers with rounded corners at the rear
  • VITON door gaskets
  • Weekly program timers
  • Activated Alumina
  • Buna-N Door gasket
  • Connection Kit
  • Mist Filter Element
  • Replacement HEPA filter
  • Silicone (red) Door gasket
  • Stainless-steel Floor Stand
  • Vacuum Pump Oil
  • White Floor Stand
All Heratherm Oven Shelf options All Vacutherm Vacuum Accessories All Linderberg/Blue M Vacuum Oven Accessories