Search
Simplify operations and experience consistently accurate results. Accelerate your scientific discovery with Thermo Scientific high-resolution accurate-mass (HRAM) mass spectrometers, which can take your laboratory’s capabilities to the next level of performance. The Thermo Scientific Orbitrap Exploris GC S Mass Spectrometer, Thermo Scientific Orbitrap Exploris GC 240 Mass Spectrometer, and the Thermo Scientific Orbitrap Exploris GC Mass Spectrometer exceed the diverse and ever-evolving needs of analytical testing and research laboratories in dioxins, pesticides, POPs, food safety, environmental, metabolomics, industrial, and pharma.
Achieve the highest confidence in compound detection through leading sensitivity, six orders of dynamic range, and high selectivity full-scan analysis. Full-scan operation can be performed up to 240,000 resolution (at m/z 200) with sub-1 ppm mass accuracy and full-scan sensitivity down to part-per-trillion concentrations. This capability drives broad-scope screening and quantitative methods for complex matrices where confident ultra-trace-level detection is required.
High-selectivity full-scan analysis offers new possibilities for high-throughput contaminant detection and food research, including easy analytical set-up, almost unlimited scope, efficient automatic data processing and retrospective data analysis. The Orbitrap Exploris GC mass spectrometer has the quantitative power of a GC triple quadrupole MS combined with the full-scan HRAM capability of Thermo Scientific Orbitrap mass analyzer technology for greater lab productivity.
Whether you need to ensure final products being shipped meet quality control standards, develop new products, or identify a suspect contaminant, high-resolution gas chromatography separations and highly sensitive HRAM acquisitions combine with intelligent identification software to quickly answer questions about the composition of each sample you inject.
The systems can be paired with Thermo Scientific TraceFinder or Thermo Scientific Compound Discoverer software, which can use multiple nominal or exact mass libraries for automatic compound identification. The libraries can be existing spectral libraries developed in-house, commercially available libraries, such as NIST or Wiley, or official Orbitrap GC-MS libraries. A more restricted library can be configured to narrow the scope of non-targeted analysis or larger libraries or combinations of libraries can be used to broaden the scope.
Be ready to run samples in minutes and confident that the system is performing at its best with automated tuning and calibration. With high-selectivity full scan analysis you will no longer have to worry about method settings. Simply acquire and process your data to answer a variety of analytical questions.
Improve lab efficiency, reduce costs and simplify operations by combining multiple targeted methods onto a single instrument. Method consolidation enables streamlined data processing and offers the flexibility to adapt to changing demands.
Experience confidence in results, sensitivity, wide dynamic range, and robustness for dioxins, pesticides, and POPs testing.
Accelerate scientific discovery with the highest-resolution GC-HRAM MS performance for advanced research, non-targeted analysis, and confident characterization of complex samples.
Transform your analytical testing workflows with high-confidence quantitation with full-scan HRAM data, simplified operation, and consistently accurate results for routine and high-throughput laboratories.
Experience the system that addresses evolving requirements for compound identification and quantitation. Go inside the instrument and view key features and capabilities.
Ten years of leading discovery with GC Orbitrap MS.
Learn about the performance you can expect from the Orbitrap Exploris GC 240 mass spectrometer.
Learn what the Orbitrap Exploris GC mass spectrometer can bring to your laboratory.
Dominic Roberts, Product Marketing Manager, Thermo Fisher Scientific
Learn how the Orbitrap Exploris GC 240 delivers high mass resolving power and provides a range of MS experiments from full scan through M/MS acquisitions.
Interview with Frank Neugebauer from Eurofins, Hamburg
Ten years of leading discovery with GC Orbitrap MS.
Learn about the performance you can expect from the Orbitrap Exploris GC 240 mass spectrometer.
Learn what the Orbitrap Exploris GC mass spectrometer can bring to your laboratory.
Dominic Roberts, Product Marketing Manager, Thermo Fisher Scientific
Learn how the Orbitrap Exploris GC 240 delivers high mass resolving power and provides a range of MS experiments from full scan through M/MS acquisitions.
Interview with Frank Neugebauer from Eurofins, Hamburg
The goal of this technical note is to demonstrate the optimization and performance of the Orbitrap Exploris GC mass spectrometer for the trace analysis of polychlorinated dibenzo-p-dioxins using high-resolution accurate-mass spectrometry.
The Thermo Scientific TriPlus RSH SMART Autosampler can eliminate the sample preparation bottleneck in GC and GC-MS analytical workflows.
Thermo Fisher Scientific has partnered with Owlstone Medical to deliver a workflow that provides a non-invasive and pain-free solution to breath testing.
Our gas chromatography (GC) systems and solutions can be customized to fit any laboratory workflow.
Our comprehensive portfolio of GC columns ensures that we have the right column to meet your analytical needs.
Our new EXTREVA ASE Accelerated Solvent Extractor offers walkaway automation of your sample preparation workflow, from sample extraction to in-cell cleanup to evaporation. It’s the perfect partner for your GC-MS analyses.
| Part number | Description | Price (USD) |
|---|---|---|
| BRE725537 | Orbitrap Exploris GC 240 Mass Spectrometer | Request a quote › |
| BRE725540 | Orbitrap Exploris GC Mass Spectrometer - 30,000 resolving power, no MS/MS | Request a quote › |
| BRE725541 | Orbitrap Exploris GC Mass Spectrometer - 60,000 resolving power, no MS/MS | Request a quote › |
| BRE725542 | Orbitrap Exploris GC Mass Spectrometer - 30,000 resolving power with MS/MS | Request a quote › |
| BRE725543 | Orbitrap Exploris GC Mass Spectrometer - 60,000 resolving power with MS/MS | Request a quote › |
* Required field