Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
A 1:100 dilution of MA5-45742 was sufficient for detection of Kir6.1 in 20 µg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. |Detects approximately 45kDa.
This antibody was formerly sold as clone S366-60.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. KCNJ8 is an integral membrane protein and inward-rectifier type potassium channel. KCNJ8, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-sensitive inward rectifier potassium channel 8; Inward rectifier K(+) channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel KIR6.1; Potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly-rectifying channel, subfamily J, member 8; uKATP-1
Gene Aliases: AI448900; gnite; KCNJ8; KIR6.1; slmbr; sltr; uKATP-1; UKATP1
UniProt ID: (Human) Q15842, (Mouse) P97794, (Rat) Q63664
Entrez Gene ID: (Human) 3764, (Mouse) 16523, (Rat) 25472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support