orbital shakers
As your research evolves, so do the needs of your laboratory. You need an orbital shaker series that can keep up with your growing needs. Whatever your research requirements, we have a wide range of shakers and accessories to cover any application from chemical mixing to cell therapy.
Choosing an orbital shaker by application
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose a shaker
Reliability
reliable orbital shakers
Our orbital shakers are designed to support your needs into the future with durable drive mechanisms backed by an industry leading warranty.
Simplicity
easy-to-use orbital shaker
Easy to use and easy to clean, our orbital shakers bring exceptional value to your lab, freeing you to move science forward.
Flexibility
flexible orbital shakers
A full offering of sizes, temperature ranges, and accessories make our orbital shakers capable of meeting your lab's growing needs.
Quick links
Choose an orbital shaker by temperature
Our orbital shakers offer a wide range of temperature capabilities to meet specific growth requirements from refrigerated models that go down to 4ºC  to high-temperature units that go up to 80ºC for growth of thermophiles.
Open air shakers
  • 0°C to 40°C
  • Use on the benchtop in ambient conditions or in a refrigerator or cold room
  • Place inside an incubator or environmental chamber
Incubated shakers
  • 5°C above ambient to 80°C
  • Incubate microorganisms or cells derived from tissue
  • Use in life science, medical research, environmental science or diagnostics
  • Ideal for production, including cell products like DNA, RNA, proteins and antibodies
Incubated and refrigerated shakers
  • 15°C below ambient to 80°C
  • Use for temperature-induced induction procedures where ambient conditions are too warm to control with an incubated shaker
  • Ideal for liquid cell cultures of insect, fish, bacteria or plant cells
Choose an orbital shaker by size
Our full range of shakers extend from small benchtop units to three-high, large stackable units, and a variety of sizes in between to accommodate a wide range of vessel types from Erlenmeyer flasks, microwell plates and beakers to centrifuge tubes.
Benchtop shakers
  • Max vessel size 1L & 2L Erlenmeyer flasks
  • 11 x 13 in. & 18 x 18 in. platforms
  • Designed to address both capacity needs and space requirements
  • Personal size shakers, available in both open air and temperature controlled, offer a small footprint for common needs.
Floor model shakers
  • Max vessel size 4L & 6L Erlenmeyer flasks
  • 18 x 30 in. platforms
  • Features consistant temperature monitoring and continous 24-hour operation
  • Incubated and refrigerated temperatures plus the ability to handle heavy loads at high speeds
Stackable shakers
  • Max vessel size 2L to 6L Erlenmeyer flasks
  • 18 x 30 in. & 18 x 18 in. platforms
  • Save valuable lab space and acheive maximum flexibility
  • Durable and dynamically balanced to stack two or three high without sacrificing performance
Top
MaxQ CO2 Plus Resistant Shaker brochure
Trust your cell cultures to a proven, reliable solution
Our CO2 resistant orbital shakers provide reliable around-the-clock operation ideally suited to keep your cells flourishing within your working environment.
Open brochure    Choose a CO2 resistant shaker
Top
Key features and benefits of our orbital shakers
  Digital Shakers High Performance Digital Shakers
Speed range 15 to 500rpm 25 to 525rpm
Drive mechanism
Triple eccentric drive accommodates heavier loads, provides uniform shaking and reliable 24/7 operation Triple counter-balanced drive is made to handle full or unbalanced loads regardless of vessel placement
Motor Maintenance-free brushless DC motor Brushless, low-heat 24V DC with moisture resistant electronics
Temperature controller PID controller, ±0.1°C over entire temperature range PID controller, ±0.1°C over entire temperature range
Display View temperature, speed and time simultaneously on three individual, push button LED displays Backlit, push button LED display contains run points and set points of temperature, speed and time, viewed simultaneously
Timer Continuous timed operation from 0.1 hour to 999 hours or 0.1 minute to 999 minutes Continuous operation or timed operation from 1 minute up to 199 hours and 59 minutes
HEPA filtration N/A Evenly distributes air inside the chamber, reducing cross contamination and improving culturing conditions
Warranty 2 years labor, 5 years parts, 10 years drive mechanism parts 2 years labor, 5 years parts, 10 years drive mechanism parts
Top
Related products
Open Air Shakers
CO2 Resistant Shakers
Reusable Plasticware
CO2 Incubators
Resources
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.