Search
Genetic variation analysis of genomic DNA (gDNA) targets via real-time PCR requires a master mix that has the utmost specificity. Applied Biosystems TaqPath ProAmp Master Mix has been specifically formulated for genetic variation analysis, enabling differentiation down to a single nucleotide change, all the while providing you with accurate and reliable results. Use TaqPath ProAmp Master mix for applications such as SNP (single nucleotide polymorphism) genotyping and CNV (copy number variation) analysis.
![]() |
|
| Check price or order now | |
Key applications |
SNP genotyping CNV analysis |
Target |
gDNA or ExtractAll crude lysate |
Concentration |
2X |
Multiplexing |
Available in 2 versions: With ROX: up to 3 targets With Mustang Purple: Up to 4 targets |
Cycling mode/run time |
Standard: ~50 min Fast: ~40 min |
Carryover contamination control |
With UNG |
Assays for SNP genotyping |
|
Assays for copy number variation analysis |
|
|
For High Resolution Melting (HRM) analysis applications such as mutation scanning and methylation analysis, use MeltDoctor HRM Master Mix.
MeltDoctor HRM Master Mix is for Research Use Only. Not for Use in diagnostic procedures.
*TaqPath master mixes are general purpose reagents labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.