Search
TaqMan Custom DNA Controls contain a linearized multi-target plasmid pool used for microbial detection research using TaqMan OpenArray plates or TaqMan array cards. These controls can be included as a positive control for panel-specific amplification, and the high concentration (5 x 107 copies/µL) products can also be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values). During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and help with troubleshooting.
| Cat. no. | Description | # of targets | Concentration (copies/µL) | Volume (µL) | List price |
|---|---|---|---|---|---|
| A50319 | TaqMan Custom DNA Control | <=20 | 1 x 105 | 1000 | $1,654 |
| A50375 | TaqMan Custom DNA Control | 21 to 30 | 1 x 105 | 1000 | $2,205 |
| A50376 | TaqMan Custom DNA Control | 31 to 50 | 1 x 105 | 1000 | $3,305 |
| A50377 | TaqMan Custom DNA Control | >50 | 1 x 105 | 1000 | Quote only |
| A50378 | TaqMan Custom DNA Control | <=20 | 5 x 107 | 50 | $2,755 |
| A50379 | TaqMan Custom DNA Control | 21 to 30 | 5 x 107 | 50 | $3,305 |
| A50380 | TaqMan Custom DNA Control | 31 to 50 | 5 x 107 | 50 | $4,410 |
| A50381 | TaqMan Custom DNA Control | >50 | 5 x 107 | 50 | Quote only |
All TaqMan Custom DNA Control products should be stored at –15°C to –25°C.
Please allow four weeks for your TaqMan Custom DNA Control to be manufactured.
For research use only. Not for use in diagnostic procedures.
Get custom support from GeneArt scientists and submit your order by email.
Send the complete form to geneartsupport@thermofisher.com.
This inventoried amplification control has targets relevant to many microbial profiling research applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance. Available in concentrations of 1 x 105 and 5 x 107 copies/µL. The higher concentration can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values).
TrueMark Respiratory Research Panels are analytically sensitive, real-time PCR syndromic panels for the analysis of a wide range of respiratory pathogens including viruses, bacteria, and fungi.
Technical support
Do you need technical support or training? We can help.
For Research Use Only. Not for use in diagnostic procedures.