Search
Discrete analyzers are integrated platforms that provide two parallel measurement techniques for your samples – photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis. Thermo Scientific Gallery discrete analyzers are equipped with fully automated liquid handling, incubation, measurement calculations, and reporting. Additionally, our discrete analyzers are robust, easy to use and provide parallel measurement of up to 20 analytes. All necessary analysis steps are automated according to precise, predefined workflows, enabling sensitive detection of the analytes in a sample to the parts per billion (ppb) level. Automation reduces hands-on time and simplifies your wet chemical laboratory testing.
Combining rugged hardware, advanced intuitive software, and simple operation, Gallery discrete analyzers are built for automated high-throughput and low-cost sample analysis workflows, minimizing manual handling errors for reliable and reproducible results. The automated workflows require no user intervention once the samples and premixed, ready-to-use reagents are loaded into the instrument. Automation not only improves the reliability and sensitivity of results compared to manual wet chemistry techniques, but it also boosts laboratory productivity by freeing staff to walk away and work on other tasks. Simply load the samples, insert the vials and leave the analyzer to do its work for up to three hours of walkaway time.
Gallery discrete analyzers offer:
These fully automated, random access, multiparameter analyzers are versatile and easy to use. They can perform up to 350 photometric tests per hour.
These fully automated, high-throughput analyzers include the precise temperature controls and flexible method parameters required for enzyme analysis.
Powered by software specifically designed for water and nutrient analysis, Thermo Scientific Gallery Aqua Master discrete analyzers help you meet local regulations.
Get all the benefits of a Gallery Plus discrete analyzer system, with the added capability of parallel bitterness testing for beer analysis.
Combine discrete analysis with ion chromatography for high-throughput, comprehensive, water and ion analysis with no method changeover required.
These ready-to-use Gallery reagents and enzymatic kits save time when analyzing water, wastewater, soil, effluents, industrial process water and more. Our kits include vials barcoded with vital identification information, such as lot number and expiration date.
Save time and reduce errors using Gallery system reagents, specially developed for food and beverage analysis. We offer more than 50 ready-to-use reagents with automated dilutions to make it easy to apply methods for a wide range of analyte concentrations.
Gallery systems use low-cost, disposable DECACELL cuvettes. A single set of cuvettes includes 10 reaction cells, mounted together for true discrete analysis. They eliminate sample carryover and the need for washing, increasing throughput, eliminating contamination and ensuring reliable results. They also require smaller (2–240 µL) sample and reagent volumes, reducing waste generation, disposal costs and, ultimately, reducing cost per analysis up to 20x.
Dedicated to analysis of beer, malt, and wort, the Gallery Plus Beermaster discrete analyzer automates labor-intensive and time-consuming tests.
Thermo Scientific Gallery discrete analyzers make wine analysis simple, accurate and reliable. Our ready-to-use system reagents are specifically developed for rapid and cost-effective analysis of juice and wine.
Learn how our discrete analyzers help you comply with the U.S. EPA Clean Water Act (CWA) by automating photometric and electrochemical analysis to provide fast, reproducible results.
To eliminate corrosion and scaling that can damage valuable industrial components and to ensure regulatory compliance, reliable water analysis must be performed continuously.
Automation of enzyme assays is becoming increasingly important and the new Gallery Enzyme Master satisfy new method development to routine QA/QC tests.
Learn about the top 10 features to consider when choosing the right discrete analyzer for your lab.
Meet your new lab partner. Gallery and Gallery Plus discrete analyzers offer faster, easier and safer chemical analysis for your laboratory.
Enable your Gallery system to perform photometric testing and automated electrochemical measurement of pH and conductivity by adding an ECM module.
Gallery discrete analyzers save you time by automating labor-intensive, time-consuming multiparameter wet chemical analysis.
Browse our searchable database of on-demand webinars about discrete analysis to find methods and applications to accelerate the work in your lab.
Visit our filterable and searchable resources library for product information, application notes, case studies, videos and more about our discrete analyzers.