Search
Primary XPS region: Te3d
Overlapping regions: Cr2p, Ag3p3/2
Binding energies of common chemical states:
| Chemical state | Binding energy Te3d5/2 |
|---|---|
| Te metal | 573.0 eV |
| Te oxide | 576.3 eV |
Native oxide charge referenced to metal peak.
Experimental information
Interpretation of XPS spectra
About this element
Symbol: Te
Date of discovery: 1782
Name origin: Greek tellus
Appearance: silverish
Discoverer: Franz Muller von Reichenstein
Obtained from: lead, copper refinement
Melting point: 722.65 K
Boiling point: 1262.95 K
Density[kg/m3]: 6240
Molar volume: 20.46 × 10-6 m3/mol
Protons/Electrons: 52
Neutrons: 76
Shell structure: 2,8,18,18,6
Electron configuration: [Kr]4d105s25p4
Oxidation state: ±2,4,6
Crystal structure: hexagonal
F. J. Muller von Reichenstein, a Romanian mining official, discovered the silver-white metalloid tellurium in 1782. While being found free in nature, tellurium is most frequently found as the telluride of gold (calaverite) and several other ores. In crystalline form, tellurium has a silvery white color and, in its pure state, the metalloid transforms to having a metallic shimmer. Similar to selenium and sulfur, the conductivity of tellurium increases slightly with exposure to light. Tellurium is combined to lead to increase its resistance to sulfuric acid and added to stainless steel and copper to make easier to machine and mill. While forming many compounds, tellurium produces none that are commercially important.
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。