We offer a wide selection of UltraClean™ surfactant-free latex microspheres—also referred to as latex beads—for research and commercial applications. High activity, superactive, and specialty latexes are available as standard products, or we can tailor particles to your specifications.
IDC Surfactant-free Latex Beads
IDC Surfactant-free Latex Beads
Latex beads are colloidal particles typically made of polystyrene. They can be made in different sizes and surface functionalities. Check out the links below for background information on the properties of polystyrene beads and how to select the right microsphere for your research.
Learn more:
High-activity Latex Beads
These UltraClean™ latex beads are hydrophobic and surfactant-free. They are stabilized against aggregation by covalently linked charge groups (sulfate, carboxyl, amidine) and have ~95% of their surface available for passive adsorption of proteins. Your proteins won’t compete with surfactants for adsorption!
|
Super Active Latex Beads
These Ultraclean™ latex beads are hydrophilic and contain a very high density of functional groups for covalent coupling of proteins to the particles. The superactive layer is a three-dimensional layer which increases the colloid stability of the particles and provides a 'soft landing' for the proteins. There is less distortion of the protein structure than if it were physically adsorbed to a rigid surface.
|
Specialty Latexes
A variety of specialty latexes, including NIST-traceable microspheres are also available.
|
Product Listing and Custom Services
Please see our product pages for a complete listing of available latex beads. Or, have our technical experts help design a product that meets your needs—we can produce a wide range of unstained, colored, or fluorescently stained latex beads. Visit our custom orders page for more details and contact information.