TSX Core Series ULT freezers
Engineered for reliability, designed for everyday use
Thermo Scientific TSX Core Series ULT freezers deliver dependable performance for everyday use. Combining reliable temperature control, robust durability, and user-centric features, TSX Core is designed to protect your critical samples while supporting workflow efficiency. Offering a cost-effective solution without compromising on quality, these freezers help researchers focus on discoveries with confidence knowing their samples are safely stored.
TSX Core Series
ULT freezers
Key Features
1. Reliability at its "Core"
  • Manufactured excellence: Built to the highest standards at our Asheville, North Carolina site
  • Proven durability: 10k+ hours of intensive lab and field runtime testing to ensure long-term reliability
  • Proven quality: Each unit undergoes rigorous end-of-line testing for quality control
Your reassurance:
  • Complete transparency: Shipped with factory documentation and a certificate of calibration*
  • Peace of mind: Backed by our 7-year warranty** for long-term confidence
2. Built to perform, designed to protect
  • Sample safety: Engineered to ensure the highest level of sample protection
  • Adaptive cooling: V-Drive technology adjusts to activity for consistent performance
Your reassurance:
  • Rapid recovery: Quickly returns to set temperature after access
  • Performance transparency: Our brochure provides upfront key performance data for all capacity sizes
3. Essential features for everyday use
  • Universal compatibility: Supports multiple voltage options
  • User-friendly interface: 5” touchscreen for easy settings and adjustments
  • Improved accessibility: Front-facing USB port for easy data logging export
  • Remote monitoring: Connectivity-enabled for streamlined workflows
  • Customizable options: Multiple capacities, inner doors, racks, and shelves to suit your needs
Your reassurance:
  • Simple integration: Ready-to-use and shipped with necessary import and qualification documents*
  • Flexible functionality: Built to fit seamlessly into your workflow
*Shipped documents: Certificate of conformance, CE declaration of conformance, certification of calibration, QC test log sheets, and QC temperature graphs.
**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.
Ordering information
Product type
Model
number
Capacity cu. ft.
(liters)
Rack capacity
(2 inch boxes)
Electrical
Plug
Shelves
Door style
General purpose
(ENERGY STAR certified, ACT label verified)
TSX40086CA
19.3 (547)
16 (400)
100-230V
50/60 Hz
View country specific details 4 shelves
(3 adjustable)
Outer (1)
Inner (2)
TSX50086CA
24.1 (682)
20 (500)
TSX60086CA
28.8 (815)
24 (600)
TSX70086CA
33.6 (950)
28 (700)
General purpose TSX40086CA-ULT 19.3 (547) 16 (400)
TSX50086CA-ULT 24.1 (682) 20 (500)
TSX60086CA-ULT 28.8 (815) 24 (600)
*Alternate Country of Origin models available. Please contact your representative for more details.
Certifications and Verifications
ENERGY STARR certified - a government backed     symbol for energy efficiency.
cULus certified - tested to meet the safety standards of Canada and the United States by Underwriters Laboratories.
Resources
Videos
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.