Search
The Thermo Scientific ST8/Megafuge 8/SL8 Compact Centrifuge Series is designed with a small footprint for multi-laboratory settings with up to 14 rotor options to adapt with evolving clinical and research needs. The series offers consistent results and innovative technologies and is available as ventilated and refrigerated models. The wide selection of rotors supports high RCFs and the flexibility to help meet evolving requirements. Microvolume rotors process microliter tubes at up to 30,279 x g to support cell culture, molecular biology, and clinical diagnostic applications. These models can run at noise levels as low as 52 dBA for a minimally disruptive lab environment.
High RCFs, small size
With dimensions as small as 12.2 x 14.6 x 18.9 in. (31 x 37 x 48 cm), our compact benchtop centrifuges can fit into especially tight lab spaces. Consider a floor model configuration, which is adjustable in height, to eliminate the need for benchtop space entirely to enable maximum flexibility and portability.
Bright, large display
The bright, large display on compact benchtop centrifuges is easy to read, even across the lab. The control panel displays instructions on programming, run conditions, alerts, and service messages.
| Sorvall brand | Megafuge brand | SL brand |
|
|
|
| Sorvall ST8 (ventilated benchtop) | Megafuge 8 (ventilated benchtop) |
SL8 (ventilated benchtop) |
| Sorvall ST8R (refrigerated benchtop) | Megafuge 8R (refrigerated benchtop) | SL8R (refrigerated benchtop) |
Sorvall ST8FR (refrigerated floor-standing) |
- |
- |
1 Certified by the National Institute for Health Protection, Porton Down, UK or TÜV NORD CERT GmbH, Germany
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.