Search
Achieving reproducible and scalable T cell activation can be difficult due to donor variability. Manual isolation steps can influence performance. These factors can affect timing control and create variation in downstream work. Gibco CTS cell therapy reagents allow for more flexible activation approaches. The Gibco CTS Detachable Dynabeads Release Buffer offers active bead detachment to adjust activation timing, while the Gibco CTS Soluble DynActivator CD3/CD28 Reagent supports bead-free activation that can occur independently of isolation steps. Both support T cell activation from development through manufacturing.
CTS T cell reagents helps manage timing, bead removal, and activation duration, supporting consistent activation. These product formats fit within closed, automated environments, bridging activation steps to later stages of the workflow. These solutions can be used in autologous and allogeneic settings to support flexible process design.
Isolation and activation steps establish the conditions that influence cell quality as they progress toward expansion. CTS cell therapy reagents are designed to support these transitions within closed or automated unit operations, enabling management of bead-based or bead-free activation with greater control.
| CTS Detachable Dynabeads Release Buffer | CTS Soluble DynActivator CD3/CD28 Reagent |
|
|
The strength and duration of CD3/CD28 activation directly influence T cell phenotype, viability, and downstream function. Both activation formats offer ways to manage these parameters across diverse starting materials, supporting efforts to maintain desired T cell subsets as cells move into expansion.
| CTS Detachable Dynabeads Release Buffer | CTS Soluble DynActivator CD3/CD28 Reagent |
Controlled bead detachment allows users to adjust activation exposure within a closed workflow that helps preserve target phenotypes through early culture. |
This bead-free option offers controlled activation that can be adjusted through concentration and exposure time. Reagent removal via simple washing or medium exchange, such as on the CTS Rotea Counterflow Centrifugation System, supports flexibility in the manufacturing process. |
Integrate Dynabeads cell separation technology with cell therapy products to support consistent handling from isolation through final processing.
CTS cell therapy reagents support the integral isolation and activation steps of T cell therapy workflows. They are available in both bead-based and bead-free formats, and can be paired with closed, automated systems, such as the CTS DynaCellect Magnetic Separation System for cell isolation and the CTS Rotea Counterflow Centrifugation System for washing and volume reduction.
CTS Detachable Dynabeads Release Buffer uses a biotin-based competitive release mechanism to actively detach CTS Detachable Dynabeads CD3/CD28 beads at your selected timepoint. Excess biotin in the buffer competes with the bead-cell interaction, allowing users to release beads rather than rely on passive dissociation. This approach supports modulation of CD3/CD28 activation strength and duration in both manual and automated workflows. It offers greater flexibility in timing and controlling activation conditions across cell therapy manufacturing steps.
CTS Soluble DynActivator CD3/CD28 Reagent is a soluble reagent that is removed through routine medium exchange or simple washing once the activation window is met. For teams working with closed or larger-scale formats, the CTS Rotea Centrifugation System can handle washing and buffer exchange in a controlled setup. These options make it simple to set defined activation times and move easily into downstream culture or engineering steps.
CTS Detachable Dynabeads Release Buffer should be stored at 2–8°C and kept protected from light. CTS Soluble DynActivator CD3/CD28 Reagent ships frozen and is stored at –15°C to –25°C until use. Each reagent for cell therapy should be handled using standard laboratory protective measures and mixed gently to maintain product integrity. Following these storage and handling guidelines helps keep the reagents ready for use.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.