Lab Centrifuges with GreenCool Technology
Unleash the power of green with every spin
Thermo Scientific Centrifuges with GreenCool Technology are Greener by Design and bring outstanding power and enhanced sustainability to the laboratory. Equipped with a next-generation cooling system, these centrifuges exemplify technological leadership and innovation. 
M Series Centrifuges
Request quote for MTR (refrigerated)
C Series Compact Centrifuges
Request quote for C1T (ventilated)
Request quote for MT (ventilated) Request quote for C1TR (refrigerated)
  Request quote for C1T
(ventilated application package)
  Request quote for C1TR 
(refrigerated application package)
X2 General Purpose Centrifuges
Request quote for X2TR (tabletop)
S2 General Purpose Centrifuges
Request quote for S2TR (tabletop)
Request quote for X2T (tabletop) Request quote for S2T (tabletop)
LYNX Superspeed Centrifuges
Request quote
Cryofuge Blood Banking Centrifuges
Request quote
BIOS Bioprocessing Centrifuges
Request quote
The advanced cooling system complies with the F-Gas Regulation (EU) 2024/573 and US EPA regulations, which aim to reduce the emission of fluorinated greenhouse gases to mitigate their impact on climate change. Fluorinated gases, including hydrofluorocarbons (HFCs), perfluorocarbons (PFCs), and sulfur hexafluoride (SF6), have a high global warming potential (GWP). These regulations seek to limit the use, production, and release of these gases.
GreenCool Technology
Less hazardous
More energy efficient
Thermo Scientific GreenCool Technology for general purpose centrifuges operates with HC (R290) with a Global Warming Potential (GWP) of 3, an immense reduction compared to previous cooling systems with a GWP of 1,430.
For floor model centrifuges, GreenCool technology utilizes carbon dioxide (CO2), a natural gas with a Global Warming Potential of 1, lower than other refrigerants.
Both R290 and CO₂ are more sustainable refrigerants that do not deplete the ozone layer.
Beyond the F-Gas compliant technology, the centrifuges offer reduced energy consumption and excellent temperature stability, helping ensure the integrity of samples as well as precise and efficient results. By leveraging application-specific solutions, these models unleash the full potential of a greener technology for top performance with every spin.
Beyond the F-Gas compliant technology, the centrifuges offer reduced energy consumption and excellent temperature stability, helping ensure the integrity of samples as well as precise and efficient results. By leveraging application-specific solutions, these models unleash the full potential of a greener technology for top performance with every spin.
M Series Centrifuges
Fast acceleration. Precision performance. Confidence in every spin.
Thermo Scientific M Series Centrifuges deliver powerful performance in a compact footprint, supporting the pace and precision of modern laboratories. Designed for reliability, safety, and ease of use, they integrate seamlessly into everyday workflows. 
Key features:
  • Compact design with powerful performance for everyday lab workflows
  • Rapid acceleration and deceleration (up to 21,100 × g in ~12 seconds) for efficient processing
  • Wide range of rotors and configurations for application flexibility
  • Intuitive controls, clear displays, and one-click lid closure for simplified operation
  • Enhanced safety with ClickSeal biocontainment lids and transparent rotor design 
GreenCool technology offers*:
  • F-Gas compliant refrigeration
  • 15% reduced energy consumption
  • Lower Global Warming Potential of 3
  • Extended life (with centrifuge recycling program in the US)
Learn more
*Compared to previous model (Sorvall Legend Micro 21R Centrifuges without GreenCool Technology).
C Series Compact Centrifuges
Compact power for separation excellence
The Thermo Scientific C Series Centrifuges are designed for clinical and research applications, offering exceptional capacity in a compact design. The centrifuges are available as ventilated or refrigerated models. The refrigerated models feature Thermo Scientific GreenCool™ Technology, a next-generation natural refrigerant cooling system for enhanced sustainability.
GreenCool technology offers*:
  • F-Gas compliant refrigeration
  • 15.7% reduced energy consumption
  • Lower Global Warming Potential of 3
  • Extended life (with centrifuge recycling program in the US)
Learn more
C Series Centrifuges combine a compact design with an intuitive interface and the flexibility to support numerous applications.
With a fixed-angle rotor RCF (Relative Centrifugal Force) up to 30,279 x g and a swinging bucket rotor RCF up to 3,260 x g, the C Series Centrifuges are capable of handling even the toughest separations. The maximum capacity of the fixed-angle rotors is 6 x 50 mL, while the swinging bucket rotors can accommodate up to 4 x 145 mL.
*Compared to previous models (Sorvall ST8/ST8R/ST8FR, Megafuge 8/8R, SL8/SL8R Centrifgues without GreenCool Technology).
X and S Series General Purpose Centrifuges
Revolving around performance
Thermo Scientific X and S General Purpose Centrifuges support efficiency across multiple applications: academic research, molecular biology, cell culture, cell and gene therapy, QA/QC laboratories and clinical diagnostics applications. Additionally, they help reduce environmental impact with a natural refrigerant cooling system.
GreenCool technology offers*:
  • F-Gas compliant refrigeration with GWP of 3
  • Up to 15% reduced energy consumption
Learn more
X and S Series General Purpose Centrifuges are designed to help provide exceptional performance, versatility and enhanced sustainability.
With fixed-angle rotor RCF (Relative Centrifugal Force) of over 25,000 × g, and swinging bucket rotor RCF of over 7,000 × g, even the toughest separations can be completed. Scientists working across biopharmaceutical, academic research and clinical diagnostic applications can choose between 4 L (4 × 1,000 mL) and 1.6 L (4x 4,00 mL) ventilated and refrigerate models. Choose from up to 18 optional rotors (X series) and up to 15 rotors (S series) to accommodate various labware, including several lightweight and corrosion-resistant Fiberlite Carbon Fiber Rotors. Additional features include Auto-Lock Rotor Exchange, ClickSeal Biocontainment Lids and optional connectivity (X series).
*Compared to previous models (Sorvall/Multifuge X Pro, Sorvall/Megafuge S Plus and SL Plus Centrifuges without GreenCool Technology).
LYNX Superspeed Centrifuges
Unleash the power of research
Thermo Scientific LYNX 4000 and 6000 Superspeed Centrifuges provide powerful performance and enhanced sustainability for research and bioprocessing laboratories.
GreenCool technology offers*:
  • F-Gas compliant refrigeration
  • 13% reduced energy consumption
  • 37% lower noise
  • 35 kg/77 lbs lighter weight
  • Reduced power demand at startup
  • Higher speeds at 4°C
LYNX centrifuges are designed for application flexibility with variety of supported labware, quick rotor exchange, up to 100,605 x g and support of even continuous flow/ zonal applications.
The LYNX 6000 centrifuge supports up to 6L capacity and 100,605 x g performance, while the LYNX 4000 centrifuge supports up to 4L capacity and 68,905 x g performance. Choose from up to 19 optional rotors to accommodate various labware, including several lightweight and corrosion-resistant Fiberlite Carbon Fiber Rotors. Additional features include Centri-Touch touchscreen interface, the Centri-Vue mobile app for real-time monitoring, GMP/GLP compliance support with Centri-Log Plus data management software and energy-efficient technologies like Smart Vacuum (LYNX 6000 model) and added safety with biocontainment lids and HEPA filter (LYNX 6000 model).
*Compared to previous models (Sorvall LYNX 4000 and 6000 Superspeed Centrifuges without GreenCool Technology).        
Cryofuge Blood Banking Centrifuges
Unleash the power of blood processing
Thermo Scientific Cryofuge 8 and 16 Blood Banking Centrifuges bring excellent productivity and enhanced sustainability to blood processing facilities.
GreenCool technology offers*:
  • F-Gas compliant refrigeration
  • 14% reduced energy consumption
  • 50% lower noise
  • 60 kg/ 132 lbs lighter weight
Cryofuge centrifuges simplify handling large volumes while helping ensure compliance with global standards.
A large capacity of up to 16 x 550 mL blood bags, user-friendly design, enhanced ergonomics and quick setup of traceable runs make these centrifuges excellent for efficient blood processing. Compliant with the Medical Device Regulation (EU) 2017/745, they help provide top performance with greener technology.
Features include Auto-Door and Auto-Lid functions for user convenience, the Centri-Touch interface for intuitive control and windshielded rotors with Eco-Spin technology for additional energy efficiency.
*Compared to previous models (Sorvall BP 8 and 16 Centrifuges/ Cryofuge 8 and 16 Centrifuges without GreenCool Technology).
BIOS Bioprocessing Centrifuges
Unleash the power of bioprocessing
Thermo Scientific BIOS A and BIOS 16 Centrifuges are designed for high-throughput bioprocessing applications and enhanced sustainability.
GreenCool technology offers*:
  • F-Gas compliant refrigeration
  • Up to 15% reduced energy consumption
  • Up to 50% lower noise
  • 60 kg/ 132 lbs lighter weight
BIOS centrifuges provide reliable performance with excellent productivity and scale-up solutions for bioprocessing applications.
With a capacity of up to 16 L (at 5,374 x g) or up to 10 L (at 12,000 x g) and a user-friendly design, the BIOS series simplifies working with large volumes while helping ensure compliance with global standards. Ergonomic features, quick setup of traceable runs and closed-system solution with CentriPAK BPC make these centrifuges excellent for bioprocessing productivity, leveraging greener technology for top performance.
Features include Auto-Door technology, full-color touchscreen interface, Auto-ID instant rotor identification and windshielded rotors with Eco-Spin technology for energy efficiency.
*Compared to previous models (Sorvall BIOS A and 16 Centrifuges without GreenCool Technology). Percentage may vary pending on model.
Made in Germany
Our Cryofuge, BIOS and LYNX centrifuge models are designed and manufactured in Osterode am Harz, Germany. This Thermo Fisher Scientific site is a certified zero-waste facility and uses 100% renewable electricity.
Resources
Video
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.