Search
| Peptide length | 6–25 amino acids |
| Manufacturing scale | ≥0.1 mg |
| Purity options | Crude (as synthesized) |
| Formulation | Suspended in 0.1% TFA in 50% (v/v) acetonitrile/water |
| Production time | 3–4 weeks |
| Modifications | Variety of modifications and labels, including heavy amino acids (isotope-labeled) |
| C-terminal residue | Lysine or Arginine |
| Counterion | Trifluoroacetic acid (TFA) |
| Delivery format | Peptides are provided in individual 2-D barcoded tubes in 96-tube plates |
| Grade 1 Fast and easy | Grade 2 Greater analysis | Grade 3 Maximum assurance | |
|---|---|---|---|
| Quality control (QC) | MS check of 5% of peptides | MS check of 100% of peptides | MS check and analysis of 100% of peptides |
| Purity | Crude | Crude | Crude |
| Peptide resynthesis | Not provided | Not provided | One resynthesis |
| Difficult peptide failed synthesis policy | You pay for entire set of peptides ordered | You only pay for peptides successfully synthesized in the requested library | You only pay for peptides successfully synthesized in the requested library |
| Included documentation | Peptide amount | Peptide amount | Peptide amount and MS Spectra |
| Minimum order | 24 peptides | 24 peptides | 24 peptides |
| Amino acid† | Code | Mass difference | Isotope | Isotopic enrichment |
|---|---|---|---|---|
| Alanine | A | +4 Da | U-13C3, 15N | >99% |
| Arginine | R | +10 Da | U-13C6, 15N4 | >99% |
| Isoleucine | I | +7 Da | U-13C6, 15N | >99% |
| Leucine | L | +7 Da | U-13C6, 15N | >99% |
| Lysine | K | +8 Da | U-13C6, 15N2 | >99% |
| Phenylalanine | F | +10 Da | U-13C9, 15N | >99% |
| Proline | P | +6 Da | U-13C5, 15N | >99% |
| Valine | V | +6 Da | U-13C5, 15N | >99% |
† Other amino acids on request.
** Others available on request.
A. The shelf life is >24 months at –20°C for lyophilized peptides protected by argon in the original non-open container. This is sequence dependent and it may be shorter for some sequences.
A. We are synthesizing our PEPotec using the solid phase Fmoc synthesis on resin. Most of our competitors use the spot technology providing poor quality peptides above 15 amino acids.
For peptide synthesis ~2.4% of the peptides submitted, due to difficult sequence were unable to be synthesized. Most of our competitors announce on their website 5%.
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design.
A. All the information required to get an accurate quote is included in the Quote Request form.
A. SRM (Simultaneous Reaction Monitoring) and MRM (Multiple Reaction Monitoring) is the terminology to describe targeted MS quantitation. SRM and MRM quantitation are typically performed on triple quadrupole instruments. PRM (Parallel Reaction Monitoring) is a new technique described by Dr. Josh Coon and Dr. Bruno Domon. PRM is used for the MS quantitation performed on Q-Exactive instruments (e.g., Orbitrap instrumentation).
| Peptide length | 6–25 amino acids |
| Manufacturing scale | ≥0.1 mg |
| Purity options | Crude (as synthesized) |
| Formulation | Suspended in 0.1% TFA in 50% (v/v) acetonitrile/water |
| Production time | 3–4 weeks |
| Modifications | Variety of modifications and labels, including heavy amino acids (isotope-labeled) |
| C-terminal residue | Lysine or Arginine |
| Counterion | Trifluoroacetic acid (TFA) |
| Delivery format | Peptides are provided in individual 2-D barcoded tubes in 96-tube plates |
| Grade 1 Fast and easy | Grade 2 Greater analysis | Grade 3 Maximum assurance | |
|---|---|---|---|
| Quality control (QC) | MS check of 5% of peptides | MS check of 100% of peptides | MS check and analysis of 100% of peptides |
| Purity | Crude | Crude | Crude |
| Peptide resynthesis | Not provided | Not provided | One resynthesis |
| Difficult peptide failed synthesis policy | You pay for entire set of peptides ordered | You only pay for peptides successfully synthesized in the requested library | You only pay for peptides successfully synthesized in the requested library |
| Included documentation | Peptide amount | Peptide amount | Peptide amount and MS Spectra |
| Minimum order | 24 peptides | 24 peptides | 24 peptides |
| Amino acid† | Code | Mass difference | Isotope | Isotopic enrichment |
|---|---|---|---|---|
| Alanine | A | +4 Da | U-13C3, 15N | >99% |
| Arginine | R | +10 Da | U-13C6, 15N4 | >99% |
| Isoleucine | I | +7 Da | U-13C6, 15N | >99% |
| Leucine | L | +7 Da | U-13C6, 15N | >99% |
| Lysine | K | +8 Da | U-13C6, 15N2 | >99% |
| Phenylalanine | F | +10 Da | U-13C9, 15N | >99% |
| Proline | P | +6 Da | U-13C5, 15N | >99% |
| Valine | V | +6 Da | U-13C5, 15N | >99% |
† Other amino acids on request.
** Others available on request.
A. The shelf life is >24 months at –20°C for lyophilized peptides protected by argon in the original non-open container. This is sequence dependent and it may be shorter for some sequences.
A. We are synthesizing our PEPotec using the solid phase Fmoc synthesis on resin. Most of our competitors use the spot technology providing poor quality peptides above 15 amino acids.
For peptide synthesis ~2.4% of the peptides submitted, due to difficult sequence were unable to be synthesized. Most of our competitors announce on their website 5%.
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design.
A. All the information required to get an accurate quote is included in the Quote Request form.
A. SRM (Simultaneous Reaction Monitoring) and MRM (Multiple Reaction Monitoring) is the terminology to describe targeted MS quantitation. SRM and MRM quantitation are typically performed on triple quadrupole instruments. PRM (Parallel Reaction Monitoring) is a new technique described by Dr. Josh Coon and Dr. Bruno Domon. PRM is used for the MS quantitation performed on Q-Exactive instruments (e.g., Orbitrap instrumentation).
For Research Use Only. Not for use in diagnostic procedures.