By using Invitrogen’s award winning BLOCK-iT RNAi Designer, you can create custom Stealth RNAi siRNA sequences for any organism. The BLOCK-iT RNAi Designer can design custom Stealth RNAi siRNA duplexes based on a cDNA sequence, GenBank accession number, or by converting your existing siRNA to a Stealth RNAi siRNA duplex.
Custom Stealth RNAi siRNA duplexes provide
  • All of the advantages of Stealth RNAi siRNA chemistry
  • Custom design of Stealth RNAi siRNA duplexes and controls for any organism
  • The ability to convert your existing siRNA sequences to Stealth RNAi siRNA
Custom design gives more flexibility
If you need a synthetic RNAi duplex to study an organism other than human, mouse or rat, Custom Stealth RNAi siRNA is the perfect option.  If you need more than 3 duplexes for your target gene, then Custom Stealth RNAi siRNA can help design additional sequences not found in the Stealth Select 3 Sets. The BLOCK-iT RNAi designer can also help design scrambled controls for each of your Custom Stealth RNAi siRNA sequences.
Ordering information
For your convenience, Custom Stealth RNAi siRNA can be ordered in 4 different synthesis scales.
Delivered AmountPriceTurnaround time 
20 nmole annealed Stealth RNAi siRNA (322µg)Contact Us≤ 7 business days 
80 nmole annealed Stealth RNAi siRNA (1.3mg)Contact Us≤ 7 business days 
1.0 µmole annealed Stealth RNAi siRNA (16.1mg)Contact Us≤ 30 business days 
1.7 µmole annealed Stealth RNAi siRNA (27.4 mg)Contact Us≤ 30 business days 
To design and order Custom Stealth RNAi siRNA, please visit the BLOCK-iT RNAi Designer. Custom Stealth RNAi siRNA can also be ordered by email.
Questions?
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments