Search
Combining the power of the QuantStudio 12K Flex Real-time PCR System with qualified TaqMan genotyping assays, this solution delivers a sample-to-answer workflow that allows you to introduce pharmacogenomic testing into your laboratory with ease.
Our SNP genotyping workflow features:
*Additional PGx data and support files at bottom of page
Our SNP genotyping solutions are available in TaqMan OpenArray (OA) and Array Plate (TAP) formats.
| TaqMan OpenArray (OA) | TaqMan Array Plates (TAP) | |
|---|---|---|
| Focus | For mid-higher throughput applications |
For low-mid throughput applications |
| Targets | TaqMan OpenArray PGx Express – 60:
|
32 assays x 12 samples: 80 plates 64 assays x 6 samples: 160 plates 128 assays x 3 samples: 320 plates |
| Use cases |
|
|
| Format and approximate workflow timeline | Off-the-Shelf and Custom 2,880 reactions with pre-spotted assays 4-hour workflow |
Off-the-Shelf and Custom 384-well plates with pre-spotted assays PGx Express (60) on 384-well plates 3-hour workflow |
| Compatible instruments |
|
|
Product |
Unit Size |
Catalog Number |
Ordering information |
1 kit |
4404846 |
||
Sizes vary |
Multiple SKUs |
||
Sizes vary |
Multiple SKUs |
We offer a simple, efficient workflow designed to optimize your process and maximize your results.
Buccal swab, saliva, or blood
Thermo Scientific KingFisher Flex system with Applied Biosystems MagMAX DNA Multi-Sample Ultra Kit
Applied Biosystems OpenArray AccuFill System
Applied Biosystems QuantStudio 12K Flex Real-Time PCR System
Drug metabolism enzyme (DME) assays run on OpenArray plates or 384-well plates
Genotyping data: In-house laboratory information systems (LIS) or relative quantification application
For Research Use Only. Not for use in diagnostic procedures.