Search
Sequence-specific DNA and RNA binding proteins (e.g. promoters, gene regulatory proteins and transcription factors) are vital in normal cellular control and in disease states such as cancer and specific genetic diseases. These regulatory proteins are potential drug targets, but as these proteins are often short-lived and scarce you need a quick and sensitive method to purify them.
Choose Streptavidin-coupled Dynabeads for a simple, quick yet reliable way to purify sequence-specific DNA or RNA binding proteins.
The high stability and binding capacity of the DNA on Dynabeads allows binding of the target proteins with kinetics similar to that of DNA in free solution
Immobilize the biotin-labelled DNA/RNA target sequence onto the beads and incubate with the cell extract. You can then isolate the bound proteins using magnetic separation. You can elute off the protein for characterization or apply the whole solid-phase complex directly e.g. to DNase footprinting studies.