Search
Thermo Scientific Amira Software provides a comprehensive toolbox for advanced 2D-5D biological characterization. It enhances time efficiency, reliability, and reproducibility with automated tools for image data analysis in life science research.
Amira Software features advanced AI-driven segmentation tools that ensure exceptional accuracy and efficiency in data analysis. Amira Software's specialized segmentation workroom combines innovative artificial intelligence with user-friendly annotation tools, streamlining the creation of ground truth data.
Amira Software provides flexible AI segmentation models, offering shallow models for rapid processing of simpler tasks and deep models for high-precision analysis of complex images. Training can be conducted in either 2D or 3D, tailored to your data needs. With its intuitive interface, superior accuracy, and efficient performance, Amira Software is an ideal solution for professionals in both research and industry.
Analyze your imaging data and obtain quantitative information:
Export your analysis and visualization work to seamlessly publish and present it:
Easily and quickly adapt Amira Software to your specific needs:
Automated image processing workflows are essential for efficient and reproducible scientific research and industrial applications.
Amira Software provides the capability to create image processing and analysis recipes in an intuitive way, allowing you to create, customize, and automate workflows, including AI-based, without any programming skills.
Save and reuse recipes for consistency and handle large datasets efficiently. You can even merge existing recipes with new recipes to increase productivity and simplify image analysis.
Amira Software's Image Recipe Designer enhances accuracy, reliability, and productivity to support your research.
Amira Software supports digital volume correlation (DVC), enabling researchers to measure non-contact, 3D volumetric displacement and strain data within biological tissues and implants under various conditions. This capability is crucial for studying the mechanical properties of bones and other structures, allowing for better understanding of strain transfer and failure mechanisms.
Amira Software features advanced AI-driven segmentation tools for precise and efficient data analysis. Its segmentation workroom incorporates innovative AI, simplifying ground truth data creation with intuitive annotation tools.
Amira Software offers versatile AI models: shallow models for speed in simple tasks and deep models for high precision in complex image analysis. Training can be done in 2D or 3D based on your data needs.
With a user-friendly interface, high accuracy, and efficient processing, Amira Software is an ideal choice for research and industry professionals, transforming images into quantifiable insights.
Amira Software provides a robust meshing engine to create simulation inputs from 3D labeled images. It generates high-quality meshes with smoothed interfaces from large multi-material segmented images, preserving thin structures and topology. You have fine control over element size distribution. Meshes can be exported to major solver formats with boundary conditions, and Amira Software supports importing solver outputs for advanced post-processing and visualization.
For Research Use Only. Not for use in diagnostic procedures.